| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006061.hg.1 |
ENST00000300190.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006061.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006061.hg.1 |
TC11000509.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006059.hg.1 |
ENST00000300190.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006059.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006059.hg.1 |
TC11000509.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006060.hg.1 |
ENST00000300190.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006060.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006060.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11006060.hg.1 |
TC11000509.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007770.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154817.hg.1 |
TC1100007770.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154818.hg.1 |
TC1100007770.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000528093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000531403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
ENST00000534071.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154820.hg.1 |
TC1100007770.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
ENST00000300190.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
ENST00000528905.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
ENST00000533885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
NM_023945.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100154821.hg.1 |
TC1100007770.hg.1 |
Clariom D TC |