| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019611.hg.1 |
ENST00000374023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019611.hg.1 |
ENST00000374026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019611.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019611.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019611.hg.1 |
TC06001659.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019609.hg.1 |
ENST00000374023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019609.hg.1 |
ENST00000374026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019609.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019609.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019609.hg.1 |
TC06001659.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008758.hg.1 |
ENST00000453101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019606.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019606.hg.1 |
ENST00000374023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019606.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019606.hg.1 |
TC06001659.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019607.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019607.hg.1 |
ENST00000374023.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019607.hg.1 |
ENST00000374026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019607.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019607.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019607.hg.1 |
TC06001659.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179518.hg.1 |
TC0600011594.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179520.hg.1 |
TC0600011594.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179512.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179512.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179512.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179512.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179512.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179512.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179512.hg.1 |
TC0600011594.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179517.hg.1 |
TC0600011594.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011594.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008758.hg.1 |
ENST00000453101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
ENST00000374021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
ENST00000374023.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
ENST00000374026.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
NM_022758.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
NM_024294.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179515.hg.1 |
TC0600011594.hg.1 |
Clariom D TC |