| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013509.hg.1 |
ENST00000300947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013509.hg.1 |
ENST00000359894.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013509.hg.1 |
ENST00000520876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013509.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013509.hg.1 |
TC19000990.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013512.hg.1 |
ENST00000300947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013512.hg.1 |
ENST00000359894.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013512.hg.1 |
ENST00000520876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013512.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013512.hg.1 |
TC19000990.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013515.hg.1 |
ENST00000300947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013515.hg.1 |
ENST00000359894.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013515.hg.1 |
ENST00000520876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013515.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013515.hg.1 |
TC19000990.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000517665.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000517665.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000517665.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000517665.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013511.hg.1 |
ENST00000300947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013511.hg.1 |
ENST00000359894.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013511.hg.1 |
ENST00000520876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013511.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013511.hg.1 |
TC19000990.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013510.hg.1 |
ENST00000300947.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013510.hg.1 |
ENST00000359894.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013510.hg.1 |
ENST00000520876.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013510.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19013510.hg.1 |
TC19000990.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
TC1900009112.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172352.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000517665.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000517665.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000517665.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000517665.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009112.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
TC1900009112.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172348.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
TC1900009112.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172350.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
TC1900009112.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172349.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
NM_024888.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
TC1900009112.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
ENST00000359894.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
ENST00000519502.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
ENST00000520876.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
NM_001270366.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900172347.hg.1 |
NM_024888.2 |
Refseq |