| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006440.hg.1 |
ENST00000351589.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006440.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006440.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006440.hg.1 |
TC22000335.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007467.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200152781.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152781.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152781.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200152781.hg.1 |
ENST00000352645.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152781.hg.1 |
ENST00000486331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200152781.hg.1 |
NM_017590.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200152781.hg.1 |
TC2200007467.hg.1 |
Clariom D TC |