| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000529268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000529268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000529268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
ENST00000529268.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026793.hg.1 |
ENST00000302251.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026793.hg.1 |
ENST00000393158.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026793.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026793.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11026793.hg.1 |
TC11002242.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100193420.hg.1 |
TC1100012155.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000302251.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000393158.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000529268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000529268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000529268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
ENST00000529268.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_025208.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012155.hg.1 |
NM_033135.3 |
Refseq |