Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00229437_m1
          See other RSG1 GE Assays ›
          Gene Symbol:
          RSG1
          Gene Name:
          REM2 and RAB like small GTPase 1
          Gene Aliases:
          C1orf89
          Chromosome Location:
          Chr.1: 16231687 - 16237445 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00229437_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RSG1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030907.3 NP_112169.2 1 - 2 537 70
          XM_011542126.2 XP_011540428.1 1 - 2 818 70
          GenBank mRNA AL559024.3 - 1 - 2 467 70
          BC002946.2 - 1 - 2 324 3955163 70
          BC008702.1 - 1 - 2 324 2966866 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013036.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002261.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RSG1

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          small GTPase mediated signal transduction
          protein transport
          regulation of exocytosis
          regulation of vesicle fusion
          cellular protein localization
          cilium assembly
          protein binding
          GTP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01034508.hg.1 TC01002261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01034508.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01034505.hg.1 TC01002261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01034505.hg.1 ENST00000375599.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034505.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01034507.hg.1 TC01002261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01034507.hg.1 ENST00000488510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034507.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01034506.hg.1 TC01002261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01034506.hg.1 ENST00000375599.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034506.hg.1 ENST00000488510.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034506.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01034503.hg.1 TC01002261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01034503.hg.1 ENST00000375599.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034503.hg.1 NM_030907.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01034504.hg.1 TC01002261.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01034504.hg.1 ENST00000375599.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01034504.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196099.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196099.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196099.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196099.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196099.hg.1 TC0100013036.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100196097.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196097.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196097.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196097.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196097.hg.1 TC0100013036.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100196098.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196098.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196098.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196098.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196098.hg.1 TC0100013036.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay TC0100013036.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196095.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196095.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196095.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196095.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196095.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196095.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196095.hg.1 TC0100013036.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100196096.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196096.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196096.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196096.hg.1 ENST00000375599.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196096.hg.1 ENST00000434014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196096.hg.1 NM_030907.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100196096.hg.1 TC0100013036.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline