| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012902.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012902.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012902.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012902.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012902.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012902.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012902.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012902.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014460.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014460.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014460.hg.1 |
TC04002941.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014464.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014464.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014464.hg.1 |
TC04002941.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014465.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014465.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014465.hg.1 |
TC04002941.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014463.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014463.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014463.hg.1 |
TC04002941.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014466.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014466.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014466.hg.1 |
TC04002941.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014462.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014462.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04014462.hg.1 |
TC04002941.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400189806.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400189806.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189806.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400189806.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189806.hg.1 |
TC0400012902.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400012902.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012902.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012902.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012902.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012902.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012902.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012902.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012902.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189804.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400189804.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189804.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400189804.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189804.hg.1 |
TC0400012902.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400189807.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400189807.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189807.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400189807.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189807.hg.1 |
TC0400012902.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400189808.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400189808.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189808.hg.1 |
ENST00000259989.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400189808.hg.1 |
NM_031950.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400189808.hg.1 |
TC0400012902.hg.1 |
Clariom D TC |