Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00232019_m1
          See other GSC2 GE Assays ›
          Gene Symbol:
          GSC2
          Gene Name:
          goosecoid homeobox 2
          Gene Aliases:
          GSCL
          Chromosome Location:
          Chr.22: 19148991 - 19150283 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00232019_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GSC2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005315.1 NP_005306.1 2 - 3 511 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007974.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000493.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GSC2

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          anatomical structure morphogenesis
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32098416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32098400 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32098425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17254702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428248 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32098420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11428241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32098423 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 894956 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P109382 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P109382 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 44701 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 55882 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03600599 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P109382 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P109382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32098416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32098400 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17254702 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32098420 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32098423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17254702 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32098400 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32098416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32098420 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32098423 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11428233 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P109382 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428239 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428247 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428233 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428237 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428242 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428246 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428234 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428241 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428243 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428238 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428236 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11428235 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254694 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254693 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254699 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254698 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254695 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11428233 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11428236 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254697 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254702 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254692 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254696 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254700 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254701 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254690 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254689 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254687 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254691 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11428234 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17254688 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11428237 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P11428235 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P109382 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03600599 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254681 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428184 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428187 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428217 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254682 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428181 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428212 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428242 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254704 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428215 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428246 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254698 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254690 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428237 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428179 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428238 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428196 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428210 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254695 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254693 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428248 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428200 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254699 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428235 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428202 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428236 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17254694 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428241 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428197 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11428219 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_005315_at - -
          Affymetrix Human Gene 1.0 ST Array 8074386 - -
          Affymetrix Human Gene 1.0 ST Array 8074387 - -
          Affymetrix Human Gene 1.0 ST Array 8074385 - -
          Affymetrix Human Gene 1.1 ST Array 8074384 - -
          Affymetrix HT HG-U133+ PM Array Plate 221441_PM_at - -
          Affymetrix Human Genome U219 Array 11759478_at - -
          Affymetrix Human Genome U219 Array 11715142_PM_s_at - -
          Affymetrix Human Genome U219 Array 11759478_PM_at - -
          Affymetrix Human Genome U219 Array 11715142_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR22008834.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008834.hg.1 NM_005315.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008834.hg.1 TC22000493.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22008832.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008832.hg.1 NM_005315.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008832.hg.1 TC22000493.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22008833.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008833.hg.1 NM_005315.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008833.hg.1 TC22000493.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200007974.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007974.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007974.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007974.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007974.hg.1 NM_005315.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007974.hg.1 NM_005315.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007974.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200156820.hg.1 TC2200007974.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200156820.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156820.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200156820.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156820.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200156822.hg.1 TC2200007974.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200156822.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156822.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200156822.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156822.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200156821.hg.1 TC2200007974.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200156821.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156821.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200156821.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156821.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007974.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007974.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007974.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007974.hg.1 ENST00000086933.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200007974.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007974.hg.1 NM_005315.1 Refseq
          Affymetrix Human Clariom D Assay TC2200007974.hg.1 NM_005315.1 Refseq
          Human Genome U133 Plus 2.0 Array 221441_at - -
          Human Genome U133 Plus 2.0 Array 221441_PM_at - -
          Human Genome U133A 2.0 Array 221441_at - -
          Human Genome U133A 2.0 Array 221441_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_005315_at - -
          AffymetrixPathwork Diagnostics Pathchip pw522885 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 221441_at9 - -
          Human Genome U133 Set 221441_PM_at - -
          Human Genome U133 Set 221441_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019460 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019460 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 122449 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_4885362-S NM_005315.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_23102-S - -
          Illumina human-6 v1.0 expression beadchip 0002970619 - -
          Illumina humanRef-8 v1.0 expression beadchip 0002970619 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 520328 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_23102 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1734951 NM_005315.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1734951 NM_005315.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zq2rp:80/100.66.77.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0