Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00234293_m1
          See other UGCG GE Assays ›
          Gene Symbol:
          UGCG
          Gene Name:
          UDP-glucose ceramide glucosyltransferase
          Gene Aliases:
          GCS, GLCT1
          Chromosome Location:
          Chr.9: 111896814 - 111935369 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00234293_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UGCG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003358.3 NP_003349.1 6 - 7 1139 68
          XM_047423844.1 XP_047279800.1 6 - 7 633 68
          GenBank mRNA AK314847.1 - 6 - 7 792 68
          BC038711.1 - 6 - 7 1188 68
          D50840.1 - 6 - 7 1027 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900008441.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000560.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Immune Tolerance Network Mammalian Gene Collection Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene UGCG

          Gene Ontology Categories:

          Function(s) Process(es)

          protein lipidation
          lipid metabolic process
          sphingolipid metabolic process
          glucosylceramide biosynthetic process
          glycosphingolipid biosynthetic process
          epidermis development
          regulation of signal transduction
          cell differentiation
          keratinocyte differentiation
          leptin-mediated signaling pathway
          neuron development
          establishment of skin barrier
          intestinal lipid absorption
          cornified envelope assembly
          protein binding
          ceramide glucosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09006258.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09006258.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09006258.hg.1 TC09000560.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000474306.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000474306.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000474306.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000489355.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000489355.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000489355.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900156436.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156436.hg.1 ENST00000474306.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156436.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900156436.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156436.hg.1 ENST00000474306.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156436.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900156436.hg.1 TC0900008441.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000474306.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000474306.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000474306.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000489355.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000489355.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000489355.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000490110.5 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 ENST00000495085.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008441.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900156437.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156437.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900156437.hg.1 ENST00000374279.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900156437.hg.1 NM_003358.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900156437.hg.1 TC0900008441.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline