| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021174.hg.1 |
ENST00000275015.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021174.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021174.hg.1 |
TC06001762.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021175.hg.1 |
ENST00000275015.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021175.hg.1 |
ENST00000477930.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021175.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021175.hg.1 |
TC06001762.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000443607.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000443607.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000443607.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000443607.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000443607.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000443607.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011913.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181928.hg.1 |
TC0600011913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181926.hg.1 |
TC0600011913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600181929.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181929.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181929.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181929.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181929.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181929.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181929.hg.1 |
TC0600011913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
ENST00000477930.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
ENST00000619360.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181927.hg.1 |
TC0600011913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600181930.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181930.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181930.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181930.hg.1 |
ENST00000275015.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181930.hg.1 |
ENST00000616555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600181930.hg.1 |
NM_004556.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600181930.hg.1 |
TC0600011913.hg.1 |
Clariom D TC |