| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000558969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000558969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000558969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000558969.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000583260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000583260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
ENST00000583260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192225.hg.1 |
TC1700011430.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000558969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000558969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000558969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000558969.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000583260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000583260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
ENST00000583260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011430.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
ENST00000006750.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
ENST00000349817.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
ENST00000392795.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
ENST00000559358.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
NM_000626.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
NM_001039933.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
NM_021602.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700192226.hg.1 |
TC1700011430.hg.1 |
Clariom D TC |