Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00241053_m1
          See other PMS2 GE Assays ›
          Gene Symbol:
          PMS2
          Gene Name:
          PMS1 homolog 2, mismatch repair system component
          Gene Aliases:
          HNPCC4, LYNCH4, MLH4, MMRCS4, PMS-2, PMS2CL, PMSL2
          Chromosome Location:
          Chr.7: 5970925 - 6009106 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00241053_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PMS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000535.7 NP_000526.2 8 - 9 935 86
          NM_001322003.2 NP_001308932.1 8 - 9 930 86
          NM_001322004.2 NP_001308933.1 7 - 8 795 86
          NM_001322005.2 NP_001308934.1 8 - 9 1125 86
          NM_001322006.2 NP_001308935.1 8 - 9 935 86
          NM_001322007.2 NP_001308936.1 7 - 8 832 86
          NM_001322008.2 NP_001308937.1 6 - 7 692 86
          NM_001322009.2 NP_001308938.1 8 - 9 1120 86
          NM_001322010.2 NP_001308939.1 7 - 8 795 86
          NM_001322011.2 NP_001308940.1 8 - 9 886 86
          NM_001322012.2 NP_001308941.1 8 - 9 881 86
          NM_001322013.2 NP_001308942.1 7 - 8 762 86
          NM_001322014.2 NP_001308943.1 8 - 9 935 86
          NM_001322015.2 NP_001308944.1 8 - 9 1105 86
          NM_001406866.1 NP_001393795.1 9 - 10 1121 86
          NM_001406868.1 NP_001393797.1 8 - 9 959 86
          NM_001406869.1 NP_001393798.1 8 - 9 827 86
          NM_001406870.1 NP_001393799.1 8 - 9 935 86
          NM_001406871.1 NP_001393800.1 8 - 9 935 86
          NM_001406872.1 NP_001393801.1 8 - 9 935 86
          NM_001406874.1 NP_001393803.1 7 - 8 767 86
          NM_001406875.1 NP_001393804.1 8 - 9 1300 86
          NM_001406876.1 NP_001393805.1 7 - 8 832 86
          NM_001406877.1 NP_001393806.1 9 - 10 1467 86
          NM_001406878.1 NP_001393807.1 8 - 9 1110 86
          NM_001406879.1 NP_001393808.1 7 - 8 970 86
          NM_001406880.1 NP_001393809.1 8 - 9 1057 86
          NM_001406881.1 NP_001393810.1 6 - 7 867 86
          NM_001406882.1 NP_001393811.1 8 - 9 1300 86
          NM_001406883.1 NP_001393812.1 7 - 8 827 86
          NM_001406884.1 NP_001393813.1 7 - 8 767 86
          NM_001406885.1 NP_001393814.1 6 - 7 599 86
          NM_001406887.1 NP_001393816.1 8 - 9 930 86
          NM_001406888.1 NP_001393817.1 8 - 9 882 86
          NM_001406889.1 NP_001393818.1 8 - 9 877 86
          NM_001406890.1 NP_001393819.1 8 - 9 925 86
          NM_001406891.1 NP_001393820.1 9 - 10 1102 86
          NM_001406892.1 NP_001393821.1 8 - 9 882 86
          NM_001406893.1 NP_001393822.1 9 - 10 1175 86
          NM_001406894.1 NP_001393823.1 8 - 9 1072 86
          NM_001406895.1 NP_001393824.1 7 - 8 742 86
          NM_001406896.1 NP_001393825.1 6 - 7 745 86
          NM_001406897.1 NP_001393826.1 8 - 9 1120 86
          NM_001406898.1 NP_001393827.1 8 - 9 1107 86
          NM_001406899.1 NP_001393828.1 9 - 10 1068 86
          NM_001406900.1 NP_001393829.1 7 - 8 1007 86
          NM_001406901.1 NP_001393830.1 7 - 8 832 86
          NM_001406902.1 NP_001393831.1 7 - 8 827 86
          NM_001406903.1 NP_001393832.1 7 - 8 832 86
          NM_001406904.1 NP_001393833.1 8 - 9 769 86
          NM_001406905.1 NP_001393834.1 8 - 9 822 86
          NM_001406906.1 NP_001393835.1 8 - 9 930 86
          NM_001406907.1 NP_001393836.1 8 - 9 882 86
          NM_001406908.1 NP_001393837.1 8 - 9 930 86
          NM_001406909.1 NP_001393838.1 7 - 8 714 86
          NM_001406910.1 NP_001393839.1 8 - 9 930 86
          NR_136154.1 8 - 9 992 86
          XM_047420482.1 XP_047276438.1 8 - 9 1206 86
          XM_047420483.1 XP_047276439.1 8 - 9 907 86
          XM_047420484.1 XP_047276440.1 8 - 9 1062 86
          GenBank mRNA AB103082.1 - 8 - 9 905 86
          AB103086.1 - 5 - 6 587 86
          AK294661.1 - 7 - 8 842 86
          AK312390.1 - 8 - 9 941 86
          BC008400.1 - 8 - 9 950 86
          BC031832.1 - 8 - 9 956 86
          BC093921.1 - 8 - 9 927 86
          BP251389.1 - 4 - 5 462 86
          PQ336165.1 - 8 - 9 905 86
          U14658.1 - 8 - 9 907 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700010182.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001124.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001322011.2, NM_001322012.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PMS2

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          mismatch repair
          cellular response to DNA damage stimulus
          response to xenobiotic stimulus
          somatic hypermutation of immunoglobulin genes
          somatic recombination of immunoglobulin gene segments
          positive regulation of isotype switching to IgA isotypes
          positive regulation of isotype switching to IgG isotypes
          nucleotide binding
          DNA binding
          single-stranded DNA binding
          nuclease activity
          endonuclease activity
          protein binding
          ATP binding
          hydrolase activity
          ATPase activity
          mismatched DNA binding
          single base insertion or deletion binding
          MutSalpha complex binding
          ATP-dependent DNA damage sensor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000265849.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000380416.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000382321.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000406569.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000415839.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000441476.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000469652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 ENST00000469652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010182.hg.1 NM_000535.6 Refseq

          Back To Top
           

          Citations (1)

          Mismatch repair and treatment resistance in ovarian cancer.
          Authors
          Helleman J, van Staveren IL, Dinjens WN, van Kuijk PF, Ritstier K, Ewing PC, van der Burg ME, Stoter G, Berns EM
          Journal
          Bmc Cancer - Page No 201 (Sun Jan 01 00:00:00 UTC 2006)
          Volume (Year)
          6 (2006)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0