Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00248373_m1
          See other TAB2 GE Assays ›
          Gene Symbol:
          TAB2
          Gene Name:
          TGF-beta activated kinase 1 (MAP3K7) binding protein 2
          Gene Aliases:
          CHTD2, MAP3K7IP2, TAB-2
          Chromosome Location:
          Chr.6: 149217926 - 149411607 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00248373_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TAB2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001292034.3 NP_001278963.1 3 - 4 1995 68
          NM_001292035.3 NP_001278964.1 3 - 4 1813 68
          NM_001369506.1 NP_001356435.1 4 - 5 2144 68
          NM_015093.6 NP_055908.1 5 - 6 2023 68
          XM_011535633.3 XP_011533935.1 5 - 6 2852 68
          XM_017010592.3 XP_016866081.1 4 - 5 1927 68
          XM_047418485.1 XP_047274441.1 3 - 4 11697 68
          XM_047418486.1 XP_047274442.1 4 - 5 1926 68
          XM_047418487.1 XP_047274443.1 4 - 5 1936 68
          XM_047418488.1 XP_047274444.1 3 - 4 1777 68
          XM_047418489.1 XP_047274445.1 3 - 4 1787 68
          XM_047418490.1 XP_047274446.1 3 - 4 1712 68
          GenBank mRNA AB018276.2 - 3 - 4 1777 68
          AF241230.1 - 2 - 3 1682 68
          AK025538.1 - 1 - 2 143 68
          AK295750.1 - 2 - 3 1636 68
          AK307650.1 - 5 - 6 2010 68
          BC035910.2 - 3 - 4 1738 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600009789.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06004102.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Human Toll-Like Receptors Pathway Human BMP Pathway Human TNF Superfamily Pathway Human Toll Pathway Core Components 1700 Inventoried Human_Toll_Pathway_Core_Components Human_Toll-Like_Receptors_Pathway Human_BMP_Pathway Human_TNF_Superfamily_Pathway Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TAB2

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          I-kappaB kinase/NF-kappaB signaling
          heart development
          response to bacterium
          negative regulation of autophagy
          response to lipopolysaccharide
          NIK/NF-kappaB signaling
          p38MAPK cascade
          defense response to bacterium
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of protein kinase activity
          protein K63-linked ubiquitination
          protein binding
          zinc ion binding
          ubiquitin binding
          metal ion binding
          binding, bridging
          K63-linked polyubiquitin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06012773.hg.1 ENST00000286332.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06012773.hg.1 ENST00000367456.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06012773.hg.1 ENST00000538427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06012773.hg.1 NM_015093.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06012773.hg.1 TC06004102.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 ENST00000367456.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 ENST00000470466.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 ENST00000538427.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 NM_001292034.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 NM_001292035.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 NM_015093.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 ENST00000367456.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 ENST00000470466.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 ENST00000538427.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 NM_001292034.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 NM_001292035.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 NM_015093.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600167065.hg.1 TC0600009789.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 199389 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline