Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00249272_m1
          See other UBE2J1 GE Assays ›
          Gene Symbol:
          UBE2J1
          Gene Name:
          ubiquitin conjugating enzyme E2 J1
          Gene Aliases:
          CGI-76, HSPC153, HSPC205, HSU93243, NCUBE-1, NCUBE1, UBC6, UBC6E, Ubc6p
          Chromosome Location:
          Chr.6: 89326625 - 89352917 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00249272_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UBE2J1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016021.2 NP_057105.2 3 - 4 568 61
          XM_011535887.1 XP_011534189.1 3 - 4 555 61
          XM_011535888.2 XP_011534190.1 3 - 4 585 61
          GenBank mRNA AF151834.1 - 3 - 4 375 61
          AF161502.1 - 3 - 4 263 61
          AJ245898.1 - 3 - 4 299 61
          AJ420578.1 - 2 - 3 217 61
          AK223464.1 - 4 - 5 307 61
          AK290574.1 - 3 - 4 410 61
          AK300629.1 - 4 - 5 313 61
          BC013973.1 - 3 - 4 530 4137664 61
          BF058850.1 - 4 - 5 487 61
          U93243.1 - 3 - 4 264 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600012542.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001940.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens HUMAN UBIQUITIN-PROTEASOME DEPENDENT PROTEOLYSIS 1700 Inventoried Human_Ubiquitin-Proteasome_Dependent_Proteolysis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene UBE2J1

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          protein ubiquitination
          protein N-linked glycosylation via asparagine
          ER-associated ubiquitin-dependent protein catabolic process
          regulation of tumor necrosis factor biosynthetic process
          negative regulation of retrograde protein transport, ER to cytosol
          ATP binding
          ubiquitin protein ligase binding
          ubiquitin protein ligase activity
          ubiquitin conjugating enzyme activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06023777.hg.1 TC06001940.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06023777.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06023777.hg.1 ENST00000536477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06023777.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay TC0600012542.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600186428.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600186428.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600186428.hg.1 ENST00000435041.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600186428.hg.1 NM_016021.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600186428.hg.1 TC0600012542.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 201805 - -
          ABI Human Genome Survey Microarray Version 2.0 211570 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline