Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00255212_m1
          See other FARS2 GE Assays ›
          Gene Symbol:
          FARS2
          Gene Name:
          phenylalanyl-tRNA synthetase 2, mitochondrial
          Gene Aliases:
          COXPD14, FARS1, HSPC320, PheRS
          Chromosome Location:
          Chr.6: 5261001 - 5771592 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00255212_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FARS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318872.1 NP_001305801.1 3 - 4 1138 115
          NM_006567.4 NP_006558.1 3 - 4 1132 115
          XM_005248812.3 XP_005248869.1 4 - 5 1196 115
          XM_006714966.2 XP_006715029.1 3 - 4 1107 115
          XM_011514247.2 XP_011512549.1 3 - 4 1107 115
          XM_011514248.2 XP_011512550.1 3 - 4 1107 115
          XM_011514249.1 XP_011512551.1 3 - 4 1107 115
          XM_011514251.2 XP_011512553.1 3 - 4 1107 115
          XM_017010186.1 XP_016865675.1 3 - 4 1915 115
          XM_017010187.1 XP_016865676.1 4 - 5 1917 115
          GenBank mRNA AF097441.1 - 2 - 3 788 115
          AK223423.1 - 3 - 4 932 115
          BC020239.1 - 3 - 4 932 5258249 115
          BC021112.1 - 3 - 4 932 5088776 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006808.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000048.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FARS2

          Panther Classification:

          Molecular Function -

          aminoacyl-tRNA synthetase

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA aminoacylation for protein translation
          phenylalanyl-tRNA aminoacylation
          tRNA processing
          tRNA binding
          magnesium ion binding
          phenylalanine-tRNA ligase activity
          protein binding
          ATP binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06000429.hg.1 ENST00000274680.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000429.hg.1 ENST00000397563.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000429.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000429.hg.1 TC06000048.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000432.hg.1 ENST00000274680.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000432.hg.1 ENST00000324331.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000432.hg.1 ENST00000397563.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000432.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000432.hg.1 TC06000048.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000431.hg.1 ENST00000274680.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000431.hg.1 ENST00000324331.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000431.hg.1 ENST00000397563.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000431.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000431.hg.1 TC06000048.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000430.hg.1 ENST00000274680.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000430.hg.1 ENST00000324331.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000430.hg.1 ENST00000397563.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000430.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000430.hg.1 TC06000048.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000428.hg.1 ENST00000274680.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000428.hg.1 ENST00000397563.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000428.hg.1 NM_006567.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000428.hg.1 TC06000048.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600146732.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146732.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146732.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146732.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146732.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146732.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146732.hg.1 TC0600006808.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146726.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146726.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146726.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146726.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146726.hg.1 TC0600006808.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146737.hg.1 TC0600006808.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay TC0600006808.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146727.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146727.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146727.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146727.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146727.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146727.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146727.hg.1 TC0600006808.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 ENST00000445533.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146735.hg.1 TC0600006808.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 ENST00000274680.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 ENST00000324331.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 ENST00000602691.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 NM_006567.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600146731.hg.1 TC0600006808.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 156192 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline