Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00255897_s1
          See other EPB41L4A-AS2 GE Assays ›
          Gene Symbol:
          EPB41L4A-AS2
          Gene Name:
          EPB41L4A antisense RNA 2 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 112419583 - 112420978 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00255897_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EPB41L4A-AS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027706.1 1 697 105
          GenBank mRNA AK002097.1 - 1 697 105
          BC107117.2 - 1 697 40023264 105
          BC107118.2 - 1 697 40023265 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500008324.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000530.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04196162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24375354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24375352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24375353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24375349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04196158 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 123742 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 2793 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P69941 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P69941 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P188975 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15473 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 8900 - -
          Agilent Human Genome CGH Microarray 44B A_14_P112696 - -
          Agilent Human Genome CGH Microarray 44A A_14_P112696 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19694 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P69941 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P69941 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00800270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375337 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375316 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375330 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375337 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196134 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375330 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24375316 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04196141 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P69941 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P188975 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P112696 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P112696 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P112696 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 68612 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P112696 - -
          Agilent Human 2 cDNA Microarray (G4101A) 4915245 - -
          Agilent Human 2 cDNA Microarray (G4101A) 4915245 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196153 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15731303 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196126 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196129 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196142 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196158 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196162 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196138 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196156 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196124 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196155 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15731306 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196151 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196140 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196145 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196160 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196132 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196144 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196149 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196134 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04196147 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15731308 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P112696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8107329 - -
          Affymetrix Human Gene 1.1 ST Array 8107328 - -
          Affymetrix HT HG-U133+ PM Array Plate 220364_PM_at - -
          Affymetrix Human Genome U219 Array 11745308_at - -
          Affymetrix Human Genome U219 Array 11745308_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR05007350.hg.1 TC05000530.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05007350.hg.1 NR_027706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008324.hg.1 ENST00000623705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008324.hg.1 ENST00000623705.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008324.hg.1 ENST00000624967.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008324.hg.1 NR_027706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008324.hg.1 NR_027706.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008324.hg.1 ENST00000623705.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008324.hg.1 ENST00000623705.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008324.hg.1 ENST00000624967.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008324.hg.1 NR_027706.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008324.hg.1 NR_027706.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500156964.hg.1 ENST00000623705.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500156964.hg.1 ENST00000624967.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500156964.hg.1 NR_027706.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500156964.hg.1 ENST00000623705.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500156964.hg.1 ENST00000624967.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500156964.hg.1 NR_027706.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500156964.hg.1 TC0500008324.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 220364_PM_at - -
          Human Genome U133 Plus 2.0 Array 220364_at - -
          Human Genome U133A 2.0 Array 220364_at - -
          Human Genome U133A 2.0 Array 220364_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw536984 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220364_at1 - -
          Human Genome U95 Set 56767_at - -
          Human Genome U95 Set 56767_PM_at - -
          Human Genome U133 Set 220364_at - -
          Human Genome U133 Set 220364_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A028622 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A028622 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 174371 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1689831 BX089466.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1689831 NR_027706.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1689831 AK002097.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1689831 BX089466.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1689831 NR_027706.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1689831 AK002097.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1689831 BX089466.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1689831 NR_027706.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1689831 AK002097.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1689831 BX089466.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1689831 NR_027706.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1689831 AK002097.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline