| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016266.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016266.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016266.hg.1 |
TC12001234.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016265.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016265.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016265.hg.1 |
TC12001234.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009921.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009921.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009921.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009921.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009921.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009921.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009921.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009921.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009921.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009921.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009921.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009921.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175014.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175014.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175014.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175014.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175014.hg.1 |
TC1200009921.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200175013.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175013.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175013.hg.1 |
ENST00000390677.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175013.hg.1 |
NM_023920.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175013.hg.1 |
TC1200009921.hg.1 |
Clariom D TC |