Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00257244_s1
          See other LOC202181 GE Assays ›
          Gene Symbol:
          LOC202181
          Gene Name:
          SUMO interacting motifs containing 1 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 177618500 - 177672277 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00257244_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC202181
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF086197.1 - 1 333 66
          AK021623.1 - 1 310 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04488452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24670166 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24670159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04488453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24670136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24670168 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 112405 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 119144 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 53534 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 42500 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 497028 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 858888 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 599848 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 24307 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 964938 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 799972 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 282001 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 799645 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 97587 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 917766 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 871324 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 583025 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 113191 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129211 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 AB006746.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 AF098642.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 BC021100.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 BC032718.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 BC070251.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 CR593697.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 CR607459.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 CR623341.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 186697 NM_021105.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 46133 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 406777 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 322370 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 219694 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 194076 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 366020 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 AK094964.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 AK124456.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 AK310762.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 BC005137.2 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 BC006182.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 CR604087.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 CR616035.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 CR626367.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 J04046.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 167321 NM_005184.2 refseq_mrna_human_NM
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P1533 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 37735 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 13599 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01402847 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01402841 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P15509715 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P15509715 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P59243615 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P1533 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P1533 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768795 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24670271 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24670247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04488455 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768788 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24670247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768788 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24670271 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768795 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768763 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15768783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04488455 - -
          Agilent Human 1B Microarray G4111A A_32_P321230 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488441 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488435 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488408 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488430 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488405 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488412 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15768655 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488417 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488439 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488426 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488446 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488454 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488428 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488421 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488451 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488452 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488415 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488431 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488443 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04488407 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01402841 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15508446 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17412195 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15509715 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01402847 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8116129 - -
          Affymetrix Human Gene 1.1 ST Array 8116128 - -
          Affymetrix HT HG-U133+ PM Array Plate 232309_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 231375_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 220609_PM_at - -
          Affymetrix Human Genome U219 Array 11757202_PM_at - -
          Affymetrix Human Genome U219 Array 11762310_x_at - -
          Affymetrix Human Genome U219 Array 11762310_PM_x_at - -
          Affymetrix Human Genome U219 Array 11757202_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500009631.hg.1 ENST00000499900.2 Ensembl
          Human Genome U133 Plus 2.0 Array 220609_at - -
          Human Genome U133 Plus 2.0 Array 232309_at - -
          Human Genome U133 Plus 2.0 Array 232309_PM_at - -
          Human Genome U133 Plus 2.0 Array 231375_PM_at - -
          Human Genome U133 Plus 2.0 Array 231375_at - -
          Human Genome U133 Plus 2.0 Array 220609_PM_at - -
          Human Genome U133A 2.0 Array 220609_PM_at - -
          Human Genome U133A 2.0 Array 220609_at - -
          AffymetrixPathwork Diagnostics Pathchip pw549966 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 232309_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220609_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 231375_at10 - -
          Human Genome U95 Set 51839_at - -
          Human Genome U95 Set 51839_PM_at - -
          Human Genome U95 Set 86804_at - -
          Human Genome U95 Set 86804_PM_at - -
          Human Genome U133 Set 220609_PM_at - -
          Human Genome U133 Set 220609_at - -
          Human Genome U133 Set 231375_at - -
          Human Genome U133 Set 232309_PM_at - -
          Human Genome U133 Set 232309_at - -
          Human Genome U133 Set 231375_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027753 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027753 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 163364 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1727331 NR_026921.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1727331 AK021623.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1719546 NR_026921.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1727331 NR_026921.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1727331 AK021623.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1719546 NR_026921.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1727331 NR_026921.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1727331 AK021623.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727331 NR_026921.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727331 AK021623.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline