Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00257509_s1
          See other AGPAT4-IT1 GE Assays ›
          Gene Symbol:
          AGPAT4-IT1
          Gene Name:
          AGPAT4 intronic transcript 1
          Gene Aliases:
          C6orf59, NCRNA00241
          Chromosome Location:
          Chr.6: 161160114 - 161161982 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00257509_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AGPAT4-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024277.1 1 702 69
          GenBank mRNA AK026765.1 - 1 711 69
          AL136708.1 - 1 556 69
          BC101668.1 - 1 178 69
          BC112291.1 - 1 178 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600013797.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06002285.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P70359 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P70359 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17146 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18604 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P70359 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P70359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215182 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215169 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215175 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420210 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215124 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215125 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215182 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215129 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215118 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215169 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215131 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215139 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215135 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05215123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25420234 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P70359 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P70359 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215178 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215184 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215176 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215169 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215125 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215146 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215142 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215129 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215167 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215109 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215153 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215113 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215183 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215132 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215157 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215173 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215165 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215133 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215136 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215137 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215155 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215162 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215150 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05215139 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8130644 - -
          Affymetrix Human Gene 1.0 ST Array 8130642 - -
          Affymetrix Human Gene 1.0 ST Array 8130643 - -
          Affymetrix Human Gene 1.1 ST Array 8130641 - -
          Affymetrix HT HG-U133+ PM Array Plate 223740_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0600013797.hg.1 ENST00000624499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013797.hg.1 NR_024277.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06028715.hg.1 TC06002285.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06028715.hg.1 NR_024277.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600195201.hg.1 ENST00000624499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600195201.hg.1 NR_024277.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600195201.hg.1 ENST00000624499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600195201.hg.1 NR_024277.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600195201.hg.1 TC0600013797.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600013797.hg.1 ENST00000624499.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600013797.hg.1 NR_024277.1 Refseq
          Human Genome U133 Plus 2.0 Array 223740_at - -
          Human Genome U133 Plus 2.0 Array 223740_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223740_at9 - -
          Human Genome U95 Set 71617_PM_at - -
          Human Genome U95 Set 71617_at - -
          Human Genome U133 Set 223740_at - -
          Human Genome U133 Set 223740_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030787 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030787 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 144531 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3241996 NR_024277.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3241996 AK026765.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241996 NR_024277.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3241996 AK026765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241996 NR_024277.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3241996 AK026765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241996 NR_024277.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3241996 AK026765.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655572 AL136708.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1655572 NR_024277.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1655572 AK026765.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1655572 AL136708.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline