Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00257547_s1
          See other ANP32A-IT1 GE Assays ›
          Gene Symbol:
          ANP32A-IT1
          Gene Name:
          ANP32A intronic transcript 1
          Gene Aliases:
          C15orf28, HsT18971, NCRNA00321
          Chromosome Location:
          Chr.15: 68803821 - 68807101 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00257547_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANP32A-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026808.1 1 410 67
          GenBank mRNA AK021784.1 - 1 410 67
          BC132845.1 - 1 70 67
          BC136847.1 - 1 70 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500009868.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001596.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P14649 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P30844 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P14649 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P378302 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 21000 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 7211 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 42137 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43179 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P14649 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P14649 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228182 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734962 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800988 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734961 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228199 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734918 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734940 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800992 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800991 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228185 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734975 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228183 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800990 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228185 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228167 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800994 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228158 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800991 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800992 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228199 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800998 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734957 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734962 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228177 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228182 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228183 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800990 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800988 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734918 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228138 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734961 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16800986 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228187 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734944 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734940 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30228189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09734975 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P30844 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P378302 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P14649 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P378302 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P14649 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734932 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734919 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734927 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16800992 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734913 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734942 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734972 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734955 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734963 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734949 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734966 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734922 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734930 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734941 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734978 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16800991 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734946 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734980 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734945 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734937 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734975 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734939 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734957 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734974 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734924 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734986 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734969 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16800999 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734961 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734915 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16800996 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16800987 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09734951 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16800994 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7990028 - -
          Affymetrix Human Gene 1.1 ST Array 7990027 - -
          Affymetrix HT HG-U133+ PM Array Plate 220710_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR15015431.hg.1 NR_026808.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15015431.hg.1 TC15001596.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500173293.hg.1 NR_026808.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500173293.hg.1 NR_026808.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500173293.hg.1 TC1500009868.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 220710_PM_at - -
          Human Genome U133 Plus 2.0 Array 220710_at - -
          Human Genome U133A 2.0 Array 220710_at - -
          Human Genome U133A 2.0 Array 220710_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw505794 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220710_at8 - -
          Human Genome U133 Set 220710_at - -
          Human Genome U133 Set 220710_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052321 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052321 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 159172 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1686662 AK021784.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1686662 NR_026808.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1686662 AK021784.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1686662 NR_026808.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1686662 AK021784.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1686662 NR_026808.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline