Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00257998_s1
          See other CPTP GE Assays ›
          Gene Symbol:
          CPTP
          Gene Name:
          ceramide-1-phosphate transfer protein
          Gene Aliases:
          GLTPD1
          Chromosome Location:
          Chr.1: 1324756 - 1328896 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00257998_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CPTP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029885.1 NP_001025056.1 3 1547 96
          XM_005244801.3 XP_005244858.1 3 1660 96
          XM_005244802.1 XP_005244859.1 2 1357 96
          XM_011542200.2 XP_011540502.1 3 1556 96
          GenBank mRNA AL122073.1 - 1 655 96
          BC004366.1 - 1 558 96
          BC098429.1 - 1 971 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100006501.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000034.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CPTP

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          glycosphingolipid metabolic process
          glycolipid transport
          ceramide 1-phosphate transport
          phospholipid binding
          phospholipid transporter activity
          glycolipid binding
          ceramide 1-phosphate binding
          ceramide 1-phosphate transporter activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 449577 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 260233 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 96770 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P225518 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P63281 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343220 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3266396 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P234105 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P63281 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P225518 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 27652 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 18946 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 7698 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53625 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 875 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 45820 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 58788 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343220 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3266396 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P225518 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P63281 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P63281 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3266396 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P225518 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3343220 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P63281 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P225518 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P234105 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P00001794 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P63281 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P225518 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P234105 - -
          Agilent Human 2 cDNA Microarray (G4101A) 1676226 - -
          Agilent Human 2 cDNA Microarray (G4101A) 1676226 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001029885_at - -
          Affymetrix Human Gene 1.0 ST Array 7896918 - -
          Affymetrix Human Gene 1.0 ST Array 8039938 - -
          Affymetrix Human Gene 1.0 ST Array 7896920 - -
          Affymetrix Human Gene 1.0 ST Array 7896919 - -
          Affymetrix Human Gene 1.1 ST Array 8039937 - -
          Affymetrix Human Gene 1.1 ST Array 7896917 - -
          Affymetrix HT HG-U133+ PM Array Plate 220734_PM_s_at - -
          Affymetrix Human Genome U219 Array 11724058_PM_x_at - -
          Affymetrix Human Genome U219 Array 11724058_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR01000432.hg.1 TC01000034.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01000432.hg.1 ENST00000343938.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01000432.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144763.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144763.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144763.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144763.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144763.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144763.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144763.hg.1 TC0100006501.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144764.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144764.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144764.hg.1 ENST00000343938.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100144764.hg.1 NM_001029885.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100144764.hg.1 TC0100006501.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 220734_s_at - -
          Human Genome U133 Plus 2.0 Array 220734_PM_s_at - -
          Human Genome U133A 2.0 Array 220734_PM_s_at - -
          Human Genome U133A 2.0 Array 220734_s_at - -
          Human Genome U95 Set 73257_PM_at - -
          Human Genome U95 Set 90503_PM_at - -
          Human Genome U95 Set 73257_at - -
          Human Genome U95 Set 90503_at - -
          Human Genome U133 Set 220734_PM_s_at - -
          Human Genome U133 Set 220734_s_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014297 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014297 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 151262 - -
          ABI Human Genome Survey Microarray Version 2.0 178690 - -
          ABI Human Genome Survey Microarray Version 2.0 215822 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2234310 CR599582.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2234310 BC098429.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2234310 AL122073.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2234310 BC004366.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2234310 NM_001029885.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 CR599582.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 BC098429.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 AL122073.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 BC004366.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 NM_001029885.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_4647-S - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_4647 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_4647 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 CR599582.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 BC098429.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 AL122073.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 BC004366.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 NM_001029885.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 CR599582.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 BC098429.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 AL122073.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 BC004366.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 NM_001029885.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline