Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00258823_s1
          See other LRRC2-AS1 GE Assays ›
          Gene Symbol:
          LRRC2-AS1
          Gene Name:
          LRRC2 antisense RNA 1
          Gene Aliases:
          LUZP3, LUZP3P, LUZPP1
          Chromosome Location:
          Chr.3: 46557398 - 46559688 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00258823_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LRRC2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_073385.1 3 113
          GenBank mRNA AI954616.1 - 1 289 113
          AJ312775.1 - 1 113
          BX279889.1 - 3 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300007264.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3795524 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P254061 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 19444 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21165 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P254061 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P254061 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8079421 - -
          Affymetrix Human Gene 1.1 ST Array 8079420 - -
          Affymetrix HT HG-U133+ PM Array Plate 236731_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 NR_073385.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 NR_073385.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007264.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300151472.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151472.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151472.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300151472.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151472.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151472.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300151472.hg.1 TC0300007264.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300151476.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151476.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151476.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300151476.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151476.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151476.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300151476.hg.1 TC0300007264.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007264.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300151474.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151474.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151474.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300151474.hg.1 ENST00000599511.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151474.hg.1 ENST00000614743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300151474.hg.1 NR_073385.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300151474.hg.1 TC0300007264.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 236731_at - -
          Human Genome U133 Plus 2.0 Array 236731_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236731_at6 - -
          Human Genome U95 Set 72451_at - -
          Human Genome U95 Set 72451_PM_at - -
          Human Genome U133 Set 236731_PM_at - -
          Human Genome U133 Set 236731_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 153224 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline