Search
Search

KRTAP4-12| Gene Symbol: KRTAP4-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031854.3 | NP_114060.1 | 1 | 688 | 133 | |
| GenBank mRNA | AJ406943.1 | - | 1 | 673 | 133 | |
| BC004180.2 | - | 1 | 653 | 133 | ||
| BC004212.2 | - | 1 | 653 | 133 | ||
| BC101159.1 | - | 1 | 665 | 133 | ||
| BC101160.1 | - | 1 | 665 | 133 | ||
| BC101162.1 | - | 1 | 665 | 133 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893148 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893145 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893144 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321409 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 135869 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P15798 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P15798 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 1949 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 46846 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P15798 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3256257 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P15798 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893148 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893142 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893145 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893148 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893142 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P15798 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321400 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321398 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321404 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321401 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321399 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321397 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P122709 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P15798 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 9316 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321398 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321406 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321404 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321400 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321408 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_031854_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8015224 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8015225 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8015228 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8015226 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8019566 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8019569 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8015227 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8019568 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8015231 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8019567 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8015229 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8019565 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8015221 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8015223 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8015230 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 224269_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11735908_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11735908_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019724.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019724.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019724.hg.1 | TC17001490.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019723.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019723.hg.1 | ENST00000455597.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019723.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019723.hg.1 | TC17001490.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019722.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019722.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019722.hg.1 | TC17001490.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019725.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019725.hg.1 | ENST00000455597.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019725.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019725.hg.1 | TC17001490.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010657.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010657.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010657.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010657.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010657.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010657.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010657.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010657.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184216.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184216.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184216.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184216.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184216.hg.1 | TC1700010657.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184214.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184214.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184214.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184214.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184214.hg.1 | TC1700010657.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184213.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184213.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184213.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184213.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184213.hg.1 | TC1700010657.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010657.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010657.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010657.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010657.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010657.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010657.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010657.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010657.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184215.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184215.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184215.hg.1 | ENST00000394014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184215.hg.1 | NM_031854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184215.hg.1 | TC1700010657.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 224269_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 224269_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224269_at9 | - | - |
| Human Genome U133 Set | 224269_at | - | - |
| Human Genome U133 Set | 224269_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008316 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008316 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 166261 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_16118236-S | BC004180.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_16118236-S | BC004212.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_16118236-S | NM_031854.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_16118236-S | AJ406943.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_2624-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0003710164 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0003710164 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 3460048 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_2624 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1698584 | BC101161.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1698584 | BC004180.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1698584 | BC004212.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1698584 | BC101159.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1698584 | BC101160.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1698584 | BC101161.1 | human_transcriptome |