Search
Search

LINC00852| Gene Symbol: LINC00852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026829.1 | 1 | 679 | 73 | ||
| GenBank mRNA | AF280797.2 | - | 1 | 679 | 73 | |
| BC015602.2 | - | 1 | 631 | 73 | ||
| BC132802.1 | - | 1 | 308 | 73 | ||
| BC132804.1 | - | 1 | 308 | 73 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 6007 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 1342 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 708483 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P251268 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 12232 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 34933 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P251268 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P251268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15411754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054990 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054988 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054983 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187374 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15411753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15411755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187374 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15411755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15411754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054988 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054983 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02054990 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15411753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22187364 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P251268 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P116447 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 96657 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P116447 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15411762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055012 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055009 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055025 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054978 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055010 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055026 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055046 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055040 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055000 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055033 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055041 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054990 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15411754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054989 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054998 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055022 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054971 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054995 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055003 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054993 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055024 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055014 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054980 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055059 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055062 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15411755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055031 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055061 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055028 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15411753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055050 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055018 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055038 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054985 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055004 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055055 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054976 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054974 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055043 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055053 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02054983 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02055057 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P116447 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8077816 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8077815 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 223725_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11744670_at | - | - |
| Affymetrix Human Genome U219 Array | 11744670_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 223725_at | - | - |
| Human Genome U133 Plus 2.0 Array | 223725_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 223725_at5 | - | - |
| Human Genome U95 Set | 46357_PM_at | - | - |
| Human Genome U95 Set | 46357_at | - | - |
| Human Genome U133 Set | 223725_at | - | - |
| Human Genome U133 Set | 223725_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022254 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A022254 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 132490 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1723254 | AF280797.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1723254 | BC015602.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1723254 | NR_026829.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1723254 | AF085991.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124236 | AF280797.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124236 | BC015602.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124236 | BC132802.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124236 | BC132804.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2124236 | NR_026829.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1723254 | AF280797.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1723254 | BC015602.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1723254 | NR_026829.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1723254 | AF085991.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1723254 | AF280797.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1723254 | BC015602.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1723254 | NR_026829.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1723254 | AF085991.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1723254 | AF280797.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1723254 | BC015602.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1723254 | NR_026829.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1723254 | AF085991.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124236 | AF280797.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124236 | BC015602.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124236 | BC132802.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124236 | BC132804.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2124236 | NR_026829.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1723254 | AF280797.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1723254 | BC015602.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1723254 | NR_026829.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1723254 | AF085991.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124236 | AF280797.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124236 | BC015602.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124236 | BC132802.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124236 | BC132804.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2124236 | NR_026829.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124236 | AF280797.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124236 | BC015602.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124236 | BC132802.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124236 | BC132804.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2124236 | NR_026829.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1723254 | AF280797.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1723254 | BC015602.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1723254 | NR_026829.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1723254 | AF085991.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124236 | AF280797.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124236 | BC015602.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124236 | BC132802.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124236 | BC132804.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2124236 | NR_026829.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1723254 | AF280797.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1723254 | BC015602.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1723254 | NR_026829.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1723254 | AF085991.1 | human_transcriptome |