Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00261831_s1
          See other LINC00260 GE Assays ›
          Gene Symbol:
          LINC00260
          Gene Name:
          long intergenic non-protein coding RNA 260
          Gene Aliases:
          C1orf217, NCRNA00260
          Chromosome Location:
          Chr.1: 203730577 - 203731853 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00261831_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00260
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046213.1 1 327 75
          GenBank mRNA BC000988.2 - 1 327 3450898 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100011269.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3210810 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P23234 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12848 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 27512 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3210810 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P23234 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P23234 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_040170_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040168_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040169_at - -
          Affymetrix Human Gene 1.0 ST Array 7908967 - -
          Affymetrix Human Gene 1.1 ST Array 7908966 - -
          Affymetrix HT HG-U133+ PM Array Plate 210711_PM_at - -
          Affymetrix Human Genome U219 Array 11760566_PM_at - -
          Affymetrix Human Genome U219 Array 11760566_at - -
          Affymetrix Human Clariom D Assay PSR0100182768.hg.1 NR_046213.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100182768.hg.1 NR_046213.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100182768.hg.1 TC0100011269.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 210711_at - -
          Human Genome U133 Plus 2.0 Array 210711_PM_at - -
          Human Genome U133A 2.0 Array 210711_PM_at - -
          Human Genome U133A 2.0 Array 210711_at - -
          AffymetrixPathwork Diagnostics Pathchip pw541257 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 210711_at4 - -
          Human Genome U133 Set 210711_PM_at - -
          Human Genome U133 Set 210711_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 105813 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1733412 BC000988.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1733412 BC000988.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1733412 BC000988.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline