Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00262115_m1
          See other LINC00525 GE Assays ›
          Gene Symbol:
          LINC00525
          Gene Name:
          long intergenic non-protein coding RNA 525
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 47761476 - 47766772 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00262115_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00525
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038407.1 2 - 3 242 72
          GenBank mRNA BC007354.2 - 2 - 3 242 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700007471.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000304.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC007354.2 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15938984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471201 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693913 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693916 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693959 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693922 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25693918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05471199 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 524806 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P120734 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P8363 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P120734 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 7379 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 10890 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16082 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P120734 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P120734 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P120734 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P8363 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P8363 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P120734 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 73295 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 56200 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01708643 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P108930 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38017826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8132709 - -
          Affymetrix Human Gene 1.0 ST Array 8132708 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553708_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR07004573.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004573.hg.1 NR_038407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004573.hg.1 TC07000304.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004576.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004576.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004576.hg.1 NR_038407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004576.hg.1 TC07000304.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07004575.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004575.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07004575.hg.1 NR_038407.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07004575.hg.1 TC07000304.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700151940.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151940.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151940.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151940.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151940.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151940.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151940.hg.1 TC0700007471.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700151943.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151943.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151943.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151943.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151943.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151943.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151943.hg.1 TC0700007471.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700151941.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151941.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151941.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151941.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151941.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151941.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151941.hg.1 TC0700007471.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700151938.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151938.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151938.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151938.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151938.hg.1 TC0700007471.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700151942.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151942.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151942.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151942.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151942.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700151942.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700151942.hg.1 TC0700007471.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000412730.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 ENST00000541385.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 NR_038407.1 Refseq
          Affymetrix Human Clariom D Assay TC0700007471.hg.1 NR_038407.1 Refseq
          Human Genome U133 Plus 2.0 Array 1553708_at - -
          Human Genome U133 Plus 2.0 Array 1553708_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553708_at6 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A034304 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A034304 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1780968 NR_038407.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1780968 BC007354.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1780968 NR_038407.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1780968 BC007354.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1780968 NR_038407.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1780968 BC007354.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1780968 NR_038407.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1780968 BC007354.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0