Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00262866_m1
          See other MGC16275 GE Assays ›
          Gene Symbol:
          MGC16275
          Gene Name:
          uncharacterized protein MGC16275
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 74209980 - 74213321 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00262866_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MGC16275
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026914.1 1 - 2 799 54
          GenBank mRNA BC043607.1 - 1 - 2 799 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700011680.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001851.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 391866 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 63712 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P315395 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P315395 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12280 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 34005 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P315395 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P315395 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31044421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31044416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17015197 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31044413 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31044426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31044416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17015197 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31044413 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460886 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31044426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460888 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460880 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10460890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31044421 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P315395 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P315395 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20727036 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03289590 - -
          Agilent Human 1B Microarray G4111A A_32_P315395 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460856 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460837 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460821 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460873 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460888 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460882 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460889 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460850 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460872 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460838 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17015194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460832 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460876 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460819 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17015197 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460890 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460844 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460836 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17015196 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17015201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460812 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460840 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460816 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460879 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460824 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460848 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460846 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460886 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460884 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460855 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460835 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460829 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10460853 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1558167_PM_a_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1558166_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR17024869.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17024869.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17024869.hg.1 TC17001851.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17024868.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17024868.hg.1 NR_026914.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17024868.hg.1 TC17001851.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700194624.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194624.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194624.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194624.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194624.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194624.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194624.hg.1 TC1700011680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194627.hg.1 TC1700011680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194629.hg.1 TC1700011680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700194626.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194626.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194626.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194626.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194626.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194626.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194626.hg.1 TC1700011680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000531617.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay TC1700011680.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194625.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194625.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194625.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194625.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194625.hg.1 TC1700011680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700194623.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194623.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194623.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194623.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194623.hg.1 TC1700011680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700194628.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194628.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194628.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194628.hg.1 ENST00000499670.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194628.hg.1 ENST00000532794.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700194628.hg.1 NR_026914.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700194628.hg.1 TC1700011680.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1558166_at - -
          Human Genome U133 Plus 2.0 Array 1558166_PM_at - -
          Human Genome U133 Plus 2.0 Array 1558167_a_at - -
          Human Genome U133 Plus 2.0 Array 1558167_PM_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558167_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558166_at8 - -
          Human Genome U95 Set 92145_at - -
          Human Genome U95 Set 92145_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008764 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008764 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 232417 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline