Search
Search

UCN2| Gene Symbol: UCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033199.3 | NP_149976.1 | 2 | 248 | 111 | |
| GenBank mRNA | AF320560.1 | - | 1 | 170 | 111 | |
| BC002647.2 | - | 2 | 248 | 3606964 | 111 | |
| BC022096.1 | - | 2 | 248 | 3916164 | 111 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22357407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02222974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22357398 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02222985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02222976 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02222980 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02222975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02222979 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 31044 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 24804 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 278556 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3279629 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P155453 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 22337 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P129361 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P129361 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29689 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 40671 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P30986581 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3279629 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3279629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357403 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357398 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222972 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15435360 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222974 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222981 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222974 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222972 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15435360 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222980 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357398 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222981 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22357403 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02222976 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P155453 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02222981 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02222976 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02222974 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02222985 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02222982 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02222983 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02222978 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02222980 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P129361 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P155453 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 114419 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 22532 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P129361 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222939 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222938 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222948 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222911 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222955 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15435361 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15435362 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222931 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222932 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222923 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15435349 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222927 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222976 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222941 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15435347 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222924 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222982 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222981 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222949 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222945 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222934 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222916 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222978 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222952 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222937 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222980 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222920 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222917 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222973 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222902 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222904 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222909 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222906 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222901 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02222946 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P129361 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_033199_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8086982 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8086984 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8086983 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8086981 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 232674_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11730718_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11730718_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013971.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013971.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013971.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013971.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013971.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013971.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024027.hg.1 | TC03003379.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024027.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024027.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024029.hg.1 | TC03003379.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024029.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024029.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024028.hg.1 | TC03003379.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024028.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024028.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024026.hg.1 | TC03003379.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024026.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024026.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013971.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013971.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013971.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013971.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013971.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013971.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207340.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207340.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207340.hg.1 | ENST00000273610.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207340.hg.1 | NM_033199.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207340.hg.1 | TC0300013971.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 232674_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 232674_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_033199_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 232674_at2 | - | - |
| Human Genome U133 Set | 232674_at | - | - |
| Human Genome U133 Set | 232674_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021031 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021031 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 198427 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_31377640-S | BC022096.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_31377640-S | BC002647.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_31377640-S | NM_033199.3 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_20791-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0002690402 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0002690402 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 240491 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_20791 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1652413 | NM_033199.3 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1652413 | BC022096.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1652413 | BC002647.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1652413 | NM_033199.3 | human_transcriptome |