| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012738.hg.1 |
ENST00000380206.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012738.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012738.hg.1 |
TC09000960.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012737.hg.1 |
ENST00000380206.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012737.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012737.hg.1 |
TC09000960.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900166409.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166409.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166409.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166409.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166409.hg.1 |
TC0900009707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900166407.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166407.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166407.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166407.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166407.hg.1 |
TC0900009707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009707.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166410.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166410.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166410.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166410.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166410.hg.1 |
TC0900009707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900166408.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166408.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166408.hg.1 |
ENST00000380206.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166408.hg.1 |
NM_000605.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166408.hg.1 |
TC0900009707.hg.1 |
Clariom D TC |