Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00265776_s1
          See other ERC2-IT1 GE Assays ›
          Gene Symbol:
          ERC2-IT1
          Gene Name:
          ERC2 intronic transcript 1
          Gene Aliases:
          C1orf1, C3orf51, Po42
          Chromosome Location:
          Chr.3: 55657206 - 55659469 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00265776_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ERC2-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024615.1 1 1446 81
          GenBank mRNA U88965.1 - 1 1357 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300011253.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001485.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P18055 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P18053 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P18055 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 38100 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43543 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4927 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P18055 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3537861 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P18055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256418 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256408 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256369 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256406 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391455 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391463 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391458 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256407 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256400 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391433 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256386 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15441515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391452 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256364 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256401 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391442 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256379 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256380 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256418 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256386 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391463 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256379 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15441515 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256369 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256380 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391455 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256408 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391431 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256364 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391442 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391458 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391452 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256401 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391433 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22391424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256406 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256400 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256407 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02256424 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P18055 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P18053 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P18053 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P18055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256397 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256390 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256380 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256434 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256398 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256371 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15441515 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256410 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256430 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256401 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256374 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256364 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256376 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256387 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256420 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256369 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15441517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256406 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256415 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256428 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256367 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256385 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256417 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256382 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256408 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256404 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256394 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256433 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256412 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15441514 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256392 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256407 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15441512 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256414 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02256423 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8088216 - -
          Affymetrix Human Gene 1.0 ST Array 8088217 - -
          Affymetrix Human Gene 1.0 ST Array 8088215 - -
          Affymetrix Human Gene 1.0 ST Array 8088218 - -
          Affymetrix Human Gene 1.0 ST Array 8088214 - -
          Affymetrix Human Gene 1.0 ST Array 8088213 - -
          Affymetrix Human Gene 1.1 ST Array 8088212 - -
          Affymetrix HT HG-U133+ PM Array Plate 208247_PM_at - -
          Affymetrix Human Genome U219 Array 11750318_PM_at - -
          Affymetrix Human Genome U219 Array 11750318_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR03026971.hg.1 TC03001485.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03026971.hg.1 NR_024615.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011253.hg.1 ENST00000629461.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300011253.hg.1 NR_024615.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300011253.hg.1 NR_024615.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300184274.hg.1 ENST00000629461.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300184274.hg.1 NR_024615.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300184274.hg.1 ENST00000629461.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300184274.hg.1 NR_024615.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300184274.hg.1 TC0300011253.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300184275.hg.1 NR_024615.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300184275.hg.1 NR_024615.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300184275.hg.1 TC0300011253.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300011253.hg.1 ENST00000629461.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300011253.hg.1 NR_024615.1 Refseq
          Affymetrix Human Clariom D Assay TC0300011253.hg.1 NR_024615.1 Refseq
          Human Genome U133 Plus 2.0 Array 208247_PM_at - -
          Human Genome U133 Plus 2.0 Array 208247_at - -
          Human Genome U133A 2.0 Array 208247_PM_at - -
          Human Genome U133A 2.0 Array 208247_at - -
          AffymetrixPathwork Diagnostics Pathchip pw553922 - -
          Human Genome U95 Set 31392_r_at - -
          Human Genome U95 Set 31392_PM_r_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208247_at4 - -
          Human Genome U133 Set 208247_PM_at - -
          Human Genome U133 Set 208247_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A021177 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A021177 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 137390 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3242335 U88965.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3242335 NR_024615.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3242335 U88965.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3242335 NR_024615.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242335 U88965.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242335 NR_024615.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3242335 U88965.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3242335 NR_024615.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline