Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00267516_s1
          See other PIGC GE Assays ›
          Gene Symbol:
          PIGC
          Gene Name:
          phosphatidylinositol glycan anchor biosynthesis class C
          Gene Aliases:
          GPI2
          Chromosome Location:
          Chr.1: 172441457 - 172444090 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00267516_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIGC
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002642.3 NP_002633.1 2 227 88
          NM_153747.1 NP_714969.1 2 204 88
          GenBank mRNA AK308201.1 - 2 183 88
          BC006539.2 - 2 174 2823399 88
          BG106700.1 - 197 88
          D85418.1 - 1 186 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100018532.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003521.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC006539.2,D85418.1,NM_002642.3,NM_153747.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PIGC

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          preassembly of GPI anchor in ER membrane
          catalytic activity
          phosphatidylinositol N-acetylglucosaminyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01054657.hg.1 TC01003521.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01054657.hg.1 ENST00000258324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054657.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054657.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054657.hg.1 ENST00000484368.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054657.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054657.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054652.hg.1 TC01003521.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01054652.hg.1 ENST00000258324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054652.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054652.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054652.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054652.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054649.hg.1 TC01003521.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01054649.hg.1 ENST00000258324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054649.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054649.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054649.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054649.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054651.hg.1 TC01003521.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01054651.hg.1 ENST00000258324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054651.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054651.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054651.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054651.hg.1 NM_153747.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054650.hg.1 TC01003521.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01054650.hg.1 ENST00000258324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054650.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054650.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01054650.hg.1 NM_002642.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01054650.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244071.hg.1 TC0100018532.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244065.hg.1 TC0100018532.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244064.hg.1 TC0100018532.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244066.hg.1 TC0100018532.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100244072.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244072.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244072.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244072.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100244072.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100244072.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100244072.hg.1 TC0100018532.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000344529.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000367728.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 ENST00000478184.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_002642.3 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_153747.1 Refseq
          Affymetrix Human Clariom D Assay TC0100018532.hg.1 NM_153747.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0