Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00267896_m1
          See other RAB3B GE Assays ›
          Gene Symbol:
          RAB3B
          Gene Name:
          RAB3B, member RAS oncogene family
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 51907956 - 51990700 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00267896_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RAB3B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002867.4 NP_002858.2 1 - 2 153 89
          GenBank mRNA AK291653.1 - 1 - 2 196 89
          AU119030.1 - 1 - 2 217 89
          BC005035.1 - 1 - 2 129 89
          M28214.1 - 1 - 2 73 88
          X13901.1 - 1 - 2 162 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014191.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002654.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC005035.1, M28214.1, NM_002867.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RAB3B

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          protein transport
          regulation of exocytosis
          antigen processing and presentation
          positive regulation of dopamine uptake involved in synaptic transmission
          regulation of vesicle size
          regulation of synaptic vesicle cycle
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          hydrolase activity
          GDP binding
          GTP-dependent protein binding
          myosin V binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01041568.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041568.hg.1 ENST00000537650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041568.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01041568.hg.1 TC01002654.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01041567.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041567.hg.1 ENST00000537650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041567.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01041567.hg.1 TC01002654.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01041569.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041569.hg.1 ENST00000537650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041569.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01041569.hg.1 TC01002654.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01041570.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041570.hg.1 ENST00000537650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01041570.hg.1 NM_002867.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01041570.hg.1 TC01002654.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100206086.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206086.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206086.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206086.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206086.hg.1 TC0100014191.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014191.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206088.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206088.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206088.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206088.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206088.hg.1 TC0100014191.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100206085.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206085.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206085.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206085.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206085.hg.1 TC0100014191.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100206087.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206087.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206087.hg.1 ENST00000371655.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206087.hg.1 NM_002867.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206087.hg.1 TC0100014191.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0