Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00269858_m1
          See other SLC7A4 GE Assays ›
          Gene Symbol:
          SLC7A4
          Gene Name:
          solute carrier family 7 member 4
          Gene Aliases:
          CAT-4, CAT4, HCAT3, VH
          Chromosome Location:
          Chr.22: 21028718 - 21032561 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00269858_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC7A4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004173.3 NP_004164.2 2 - 3 1059 82
          XM_047441472.1 XP_047297428.1 1 - 2 1737 82
          GenBank mRNA AJ000730.1 - 2 - 3 1069 82
          BC008814.2 - 2 - 3 1056 82
          BC062565.1 - 2 - 3 1050 82
          BC107160.1 - 1 - 2 1019 82
          BC107161.1 - 1 - 2 1019 82
          CR456580.1 - 1 - 2 1021 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200008069.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000529.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC7A4

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          amino acid transport
          transmembrane transport
          basic amino acid transmembrane transport
          amino acid transmembrane transporter activity
          basic amino acid transmembrane transporter activity
          transmembrane transporter activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22009657.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009657.hg.1 ENST00000403586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009657.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009657.hg.1 TC22000529.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200006687.hg.1 ENST00000450652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006687.hg.1 ENST00000450652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006687.hg.1 ENST00000450652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009659.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009659.hg.1 ENST00000403586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009659.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009659.hg.1 TC22000529.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009658.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009658.hg.1 ENST00000403586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009658.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009658.hg.1 TC22000529.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22009664.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22009664.hg.1 NM_004173.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22009664.hg.1 TC22000529.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2200006687.hg.1 ENST00000450652.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006687.hg.1 ENST00000450652.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006687.hg.1 ENST00000450652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157728.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157728.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157728.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157728.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157728.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157728.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157728.hg.1 TC2200008069.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008069.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157727.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157727.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157727.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157727.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157727.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157727.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157727.hg.1 TC2200008069.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157729.hg.1 TC2200008069.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 ENST00000382932.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 ENST00000403586.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 ENST00000426145.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 NM_004173.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200157730.hg.1 TC2200008069.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 193063 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline