Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00274247_s1
          See other CCT8L2 GE Assays ›
          Gene Symbol:
          CCT8L2
          Gene Name:
          chaperonin containing TCP1 subunit 8 like 2
          Gene Aliases:
          CESK1
          Chromosome Location:
          Chr.22: 16590751 - 16592810 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00274247_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCT8L2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014406.5 NP_055221.1 1 757 79
          GenBank mRNA BC033797.1 - 1 757 79
          BC100811.2 - 1 623 79
          BC100812.2 - 1 623 79
          BC100813.1 - 1 623 79
          BC139842.1 - 1 516 79
          CR456508.1 - 1 612 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007871.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000461.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCT8L2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          potassium ion transmembrane transport
          anion transmembrane transport
          nucleotide binding
          anion channel activity
          ATP binding
          calcium-activated potassium channel activity
          unfolded protein binding
          ATP-dependent protein folding chaperone

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32088943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17249587 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17249662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17249581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17249698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32088984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32088961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11421961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11421962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17249673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17249686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11421964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32088959 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11421960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32088963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32089015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32088936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17249693 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 849878 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 140149 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P29110 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P29110 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 22218 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20438 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P29110 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P29110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17249656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17249662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17249673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32088982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32088984 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17249656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17249673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32088982 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17249662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32088984 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P29110 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11421965 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11421963 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11421964 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P29110 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17249710 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11421964 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11421960 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_014406_at - -
          Affymetrix Human Gene 1.0 ST Array 8074198 - -
          Affymetrix Human Gene 1.0 ST Array 8074200 - -
          Affymetrix Human Gene 1.0 ST Array 8074197 - -
          Affymetrix Human Gene 1.0 ST Array 8074199 - -
          Affymetrix Human Gene 1.1 ST Array 8074196 - -
          Affymetrix HT HG-U133+ PM Array Plate 220508_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR22008337.hg.1 ENST00000359963.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008337.hg.1 NM_014406.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008337.hg.1 TC22000461.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700009739.hg.1 ENST00000465400.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008339.hg.1 ENST00000359963.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008339.hg.1 NM_014406.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008339.hg.1 TC22000461.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2200155897.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200155897.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200155897.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200155897.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200155897.hg.1 TC2200007871.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700009739.hg.1 ENST00000465400.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay TC2200007871.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200155899.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200155899.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200155899.hg.1 ENST00000359963.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2200155899.hg.1 NM_014406.4 Refseq
          Affymetrix Human Clariom D Assay PSR2200155899.hg.1 TC2200007871.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 220508_at - -
          Human Genome U133 Plus 2.0 Array 220508_PM_at - -
          Human Genome U133A 2.0 Array 220508_PM_at - -
          Human Genome U133A 2.0 Array 220508_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_014406_at - -
          AffymetrixPathwork Diagnostics Pathchip pw573087 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220508_at6 - -
          Human Genome U95 Set 74266_PM_at - -
          Human Genome U95 Set 74266_at - -
          Human Genome U133 Set 220508_at - -
          Human Genome U133 Set 220508_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019417 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019417 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 163518 - -
          ABI Human Genome Survey Microarray Version 2.0 180063 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1727272 BC139842.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1727272 CR456508.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1727272 BC100811.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1727272 BC100812.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1727272 BC100813.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1687049 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1727272 BC139842.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1727272 CR456508.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1727272 BC100811.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1727272 BC100812.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1727272 BC100813.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1727272 BC139842.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1727272 CR456508.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1727272 BC100811.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1727272 BC100812.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1727272 BC100813.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1727272 BC139842.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1727272 CR456508.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1727272 BC100811.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1727272 BC100812.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1727272 BC100813.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31560866-S BC139842.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31560866-S CR456508.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31560866-S BC100811.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31560866-S BC100812.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_31560866-S BC100813.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_8060-S - -
          Illumina human-6 v1.0 expression beadchip 0003830411 - -
          Illumina humanRef-8 v1.0 expression beadchip 0003830411 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1687049 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1687049 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 2680202 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_8060 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1727272 BC139842.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1727272 CR456508.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1727272 BC100811.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1727272 BC100812.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1727272 BC100813.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727272 BC139842.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727272 CR456508.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727272 BC100811.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727272 BC100812.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727272 BC100813.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1727272 BC139842.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1727272 CR456508.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1727272 BC100811.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1727272 BC100812.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1727272 BC100813.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1687049 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1727272 BC139842.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1727272 CR456508.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1727272 BC100811.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1727272 BC100812.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1727272 BC100813.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0