Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00289329_m1
          See other ZSCAN12P1 GE Assays ›
          Gene Symbol:
          ZSCAN12P1
          Gene Name:
          zinc finger and SCAN domain containing 12 pseudogene 1
          Gene Aliases:
          ZNF187p1, ZNF305P1, ZNF96L1, ZNF96P1, ZSCAN12L1, dJ313I6.7
          Chromosome Location:
          Chr.6: 28090807 - 28095715 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00289329_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZSCAN12P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024063.2 1 - 2 678 94
          GenBank mRNA AK055113.1 - 1 - 2 340 94
          BC040524.1 - 1 - 2 334 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007396.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000274.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 898746 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 152664 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 932747 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3422113 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P360518 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11557 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4890 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P17490215 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3422113 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3422113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821486 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631272 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821477 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821483 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821484 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631272 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821486 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821477 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631269 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821484 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821483 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24821493 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17490215 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P360518 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631260 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631286 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631196 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631262 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631284 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631210 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631199 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631288 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631306 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631203 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631268 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631213 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631292 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631198 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631209 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631310 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631276 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631294 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631265 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631259 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631255 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631201 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17490215 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 239157_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06002278.hg.1 TC06000274.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06002278.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06002278.hg.1 NR_024063.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06002279.hg.1 TC06000274.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06002279.hg.1 NR_024063.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 ENST00000406489.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06002277.hg.1 TC06000274.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06002277.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06002277.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150073.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150073.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150073.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150073.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150073.hg.1 TC0600007396.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600150076.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150076.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150076.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150076.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150076.hg.1 TC0600007396.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 ENST00000406489.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007396.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150074.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150074.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150074.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150074.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150074.hg.1 TC0600007396.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600150072.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150072.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150072.hg.1 TC0600007396.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600150075.hg.1 ENST00000406489.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150075.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150075.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150075.hg.1 ENST00000406489.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150075.hg.1 ENST00000529104.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600150075.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150075.hg.1 TC0600007396.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600150077.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150077.hg.1 NR_024063.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600150077.hg.1 TC0600007396.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 239157_PM_at - -
          Human Genome U133 Plus 2.0 Array 239157_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239157_at8 - -
          Human Genome U95 Set 50619_PM_at - -
          Human Genome U95 Set 50619_at - -
          Human Genome U95 Set 80434_PM_r_at - -
          Human Genome U95 Set 80434_r_at - -
          Human Genome U133 Set 239157_PM_at - -
          Human Genome U133 Set 239157_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031168 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031169 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031169 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031168 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 693229 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3240829 BC040524.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3240829 NR_024063.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3240829 BC040524.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3240829 NR_024063.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3240829 BC040524.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3240829 NR_024063.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3240829 BC040524.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3240829 NR_024063.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline