Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00293288_m1
          See other SLC25A44 GE Assays ›
          Gene Symbol:
          SLC25A44
          Gene Name:
          solute carrier family 25 member 44
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 156194104 - 156212796 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00293288_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC25A44
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286184.2 NP_001273113.1 3 - 4 933 74
          NM_001377385.1 NP_001364314.1 4 - 5 1094 74
          NM_001377386.1 NP_001364315.1 5 - 6 1799 74
          NM_001377387.1 NP_001364316.1 4 - 5 1638 74
          NM_001377388.1 NP_001364317.1 4 - 5 1070 74
          NM_014655.4 NP_055470.1 3 - 4 909 74
          NR_104408.2 3 - 4 432 74
          NR_104411.2 2 - 3 271 74
          XM_011510180.2 XP_011508482.1 5 - 6 1823 74
          XM_011510181.3 XP_011508483.1 4 - 5 1662 74
          XM_047435097.1 XP_047291053.1 5 - 6 2343 74
          XM_047435098.1 XP_047291054.1 5 - 6 2319 74
          XM_047435101.1 XP_047291057.1 4 - 5 1590 74
          GenBank mRNA AB007915.3 - 3 - 4 908 74
          AK074912.1 - 3 - 4 926 74
          AK075002.1 - 3 - 4 901 74
          AK294524.1 - 4 - 5 828 74
          AK299747.1 - 4 - 5 583 74
          AK307968.1 - 5 - 6 693 74
          AK309678.1 - 2 - 3 272 74
          BC008843.2 - 3 - 4 905 74
          BC039854.1 - 3 - 4 884 74
          HY053859.1 - 3 - 4 436 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010159.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001315.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC25A44

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transport
          branched-chain amino acid catabolic process
          branched-chain amino acid transport
          transmembrane transport
          regulation of cold-induced thermogenesis
          branched-chain amino acid transmembrane transporter activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000468973.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000468973.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000482737.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 ENST00000482737.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104412.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104412.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104412.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010159.hg.1 NR_104412.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01020537.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01020537.hg.1 ENST00000423538.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01020537.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01020537.hg.1 NM_014655.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01020537.hg.1 TC01001315.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 ENST00000468973.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NR_104408.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NR_104411.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NR_104412.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 ENST00000468973.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NR_104408.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NR_104411.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 NR_104412.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100174393.hg.1 TC0100010159.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000359511.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000423538.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000468973.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000468973.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000469537.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000482737.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 ENST00000482737.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_001286184.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NM_014655.3 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104408.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104411.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104412.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104412.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104412.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010159.hg.1 NR_104412.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 134864 - -
          ABI Human Genome Survey Microarray Version 2.0 219638 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0