| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02008810.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02008810.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02008810.hg.1 |
TC02000575.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158705.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158705.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158705.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158705.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158705.hg.1 |
TC0200008481.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013549.hg.1 |
ENST00000422468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
ENST00000272395.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008481.hg.1 |
NM_138800.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
ENST00000432468.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013489.hg.1 |
NM_001164464.1 |
Refseq |