Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00300261_m1
          See other DYNC2H1 GE Assays ›
          Gene Symbol:
          DYNC2H1
          Gene Name:
          dynein cytoplasmic 2 heavy chain 1
          Gene Aliases:
          ATD3, DHC1b, DHC2, DNCH2, DYH1B, SRPS2B, SRTD3, hdhc11
          Chromosome Location:
          Chr.11: 103109426 - 103479863 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00300261_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DYNC2H1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080463.2 NP_001073932.1 30 - 31 4759 59
          NM_001377.3 NP_001368.2 30 - 31 4759 59
          XM_006718903.3 XP_006718966.1 30 - 31 4759 59
          XM_017018291.2 XP_016873780.1 30 - 31 4759 59
          XM_017018292.2 XP_016873781.1 27 - 28 4008 59
          XM_017018293.2 XP_016873782.1 30 - 31 4759 59
          GenBank mRNA AB290167.1 - 30 - 31 4808 59
          U53531.1 - 9 - 10 1342 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100008927.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000963.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Homo sapiens Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          microtubule-based movement
          Golgi organization
          determination of left/right symmetry
          heart development
          dorsal/ventral pattern formation
          protein processing
          spinal cord motor neuron differentiation
          central nervous system neuron differentiation
          cell projection organization
          neuron differentiation
          embryonic limb morphogenesis
          forebrain development
          intraciliary retrograde transport
          positive regulation of smoothened signaling pathway
          cilium morphogenesis
          cilium movement involved in cell motility
          coronary vasculature development
          protein localization to cilium
          non-motile cilium assembly
          nucleotide binding
          motor activity
          protein binding
          ATP binding
          ATP-dependent microtubule motor activity, minus-end-directed
          dynein intermediate chain binding
          dynein light intermediate chain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11011962.hg.1 ENST00000375735.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11011962.hg.1 ENST00000398093.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11011962.hg.1 NM_001080463.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11011962.hg.1 NM_001377.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11011962.hg.1 TC11000963.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 ENST00000375735.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 ENST00000398093.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 NM_001080463.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 NM_001377.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 ENST00000375735.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 ENST00000398093.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 NM_001080463.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 NM_001377.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100165870.hg.1 TC1100008927.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline