Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00323583_m1
          See other HIPK1 GE Assays ›
          Gene Symbol:
          HIPK1
          Gene Name:
          homeodomain interacting protein kinase 1
          Gene Aliases:
          Myak, Nbak2
          Chromosome Location:
          Chr.1: 113929197 - 113977869 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00323583_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HIPK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152696.3 NP_689909.2 14 - 15 3172 91
          NM_181358.2 NP_852003.1 12 - 13 1892 91
          NM_198268.2 NP_938009.1 14 - 15 3172 91
          NM_198269.2 NP_938010.1 13 - 14 2083 91
          XM_005270609.3 XP_005270666.1 14 - 15 3349 91
          XM_005270610.4 XP_005270667.1 14 - 15 3346 91
          XM_005270612.4 XP_005270669.1 14 - 15 3311 91
          XM_005270613.4 XP_005270670.1 16 - 17 3262 91
          XM_006710443.3 XP_006710506.1 14 - 15 3247 91
          XM_011540977.2 XP_011539279.1 13 - 14 3229 91
          XM_017000605.1 XP_016856094.1 14 - 15 3308 91
          GenBank mRNA AB014530.1 - 5 - 6 852 91
          AB089957.1 - 14 - 15 3136 91
          AK122662.1 - 14 - 15 3170 91
          AK302726.1 - 14 - 15 3356 91
          AL833829.1 - 13 - 14 2061 91
          BC028408.1 - 13 - 14 2088 4821915 91
          BC033012.2 - 14 - 15 3172 4830408 91
          BC036057.1 - 12 - 13 1892 5272816 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100009480.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001003.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HIPK1

          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          smoothened signaling pathway
          positive regulation of cell proliferation
          anterior/posterior pattern specification
          regulation of tumor necrosis factor-mediated signaling pathway
          retina layer formation
          neuron differentiation
          adherens junction assembly
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of angiogenesis
          embryonic camera-type eye morphogenesis
          embryonic retina morphogenesis in camera-type eye
          definitive hemopoiesis
          lens induction in camera-type eye
          iris morphogenesis
          endothelial cell apoptotic process
          extrinsic apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          protein serine/threonine kinase activity
          protein binding
          ATP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 TC01001003.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000340480.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000369553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000369554.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000369555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000369558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000369559.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000369561.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000406344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000426820.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 ENST00000443627.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 NM_152696.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 NM_181358.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 NM_198268.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01016007.hg.1 NM_198269.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01016008.hg.1 TC01001003.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01016008.hg.1 ENST00000369559.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01016008.hg.1 NM_152696.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000340480.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000361587.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369553.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369555.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369558.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369559.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369561.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000426820.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000626993.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 NM_152696.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 NM_181358.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 NM_198268.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 NM_198269.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000340480.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000361587.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369553.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369555.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369558.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369559.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000369561.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000426820.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 ENST00000626993.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 NM_152696.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 NM_181358.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 NM_198268.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 NM_198269.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100168355.hg.1 TC0100009480.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100168356.hg.1 ENST00000369559.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168356.hg.1 NM_152696.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100168356.hg.1 ENST00000369559.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168356.hg.1 NM_152696.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100168356.hg.1 TC0100009480.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100168357.hg.1 ENST00000369559.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168357.hg.1 NM_152696.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100168357.hg.1 ENST00000369559.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100168357.hg.1 NM_152696.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100168357.hg.1 TC0100009480.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline