| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
236769_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563249.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563249.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563249.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000568421.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000568421.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000568421.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011219.hg.1 |
ENST00000568421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563249.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563249.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563249.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000563434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000568421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000568421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000568421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011219.hg.1 |
ENST00000568421.1 |
Ensembl |
| Human Genome U133 Plus 2.0 Array |
1559067_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
236769_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559066_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559067_PM_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
236769_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1559066_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
236769_at9 |
- |
- |
| Human Genome U95 Set |
55078_PM_at |
- |
- |
| Human Genome U95 Set |
55078_at |
- |
- |
| Human Genome U95 Set |
91225_at |
- |
- |
| Human Genome U95 Set |
91225_PM_at |
- |
- |
| Human Genome U133 Set |
236769_at |
- |
- |
| Human Genome U133 Set |
236769_PM_at |
- |
- |