Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00328135_m1
          See other LOC441178 GE Assays ›
          Gene Symbol:
          LOC441178
          Gene Name:
          uncharacterized LOC441178
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 167679626 - 167683787 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00328135_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC441178
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_117092.1 2 - 3 130 70
          GenBank mRNA AK056600.1 - 2 - 3 130 70
          AL832737.1 - 4 - 5 1442 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600013936.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 585582 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 410532 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P522786 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3309222 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P522786 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 6124 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 20839 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 27708 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3309222 - -
          Agilent Human 1B Microarray G4111A A_32_P845232 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XR_040770_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040769_at - -
          Affymetrix Human Gene 1.0 ST Array XR_040768_at - -
          Affymetrix Human Gene 1.0 ST Array 8130981 - -
          Affymetrix Human Gene 1.0 ST Array 8130808 - -
          Affymetrix Human Gene 1.0 ST Array 8130810 - -
          Affymetrix Human Gene 1.0 ST Array 8130809 - -
          Affymetrix Human Gene 1.1 ST Array 8130807 - -
          Affymetrix Human Gene 1.1 ST Array 8130980 - -
          Affymetrix HT HG-U133+ PM Array Plate 1563894_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1561685_PM_a_at - -
          Affymetrix Human Genome U219 Array 11745412_PM_s_at - -
          Affymetrix Human Genome U219 Array 11745412_s_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0600013936.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013936.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013936.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013936.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600013936.hg.1 NR_117092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013936.hg.1 NR_117092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013936.hg.1 NR_117092.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600013936.hg.1 NR_117092.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196014.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196014.hg.1 NR_117092.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196014.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196014.hg.1 NR_117092.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196014.hg.1 TC0600013936.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600196015.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196015.hg.1 NR_117092.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196015.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600196015.hg.1 NR_117092.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600196015.hg.1 TC0600013936.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600013936.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013936.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013936.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013936.hg.1 ENST00000400831.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600013936.hg.1 NR_117092.1 Refseq
          Affymetrix Human Clariom D Assay TC0600013936.hg.1 NR_117092.1 Refseq
          Affymetrix Human Clariom D Assay TC0600013936.hg.1 NR_117092.1 Refseq
          Affymetrix Human Clariom D Assay TC0600013936.hg.1 NR_117092.1 Refseq
          Human Genome U133 Plus 2.0 Array 1563894_at - -
          Human Genome U133 Plus 2.0 Array 1563894_PM_at - -
          Human Genome U133 Plus 2.0 Array 1561685_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1561685_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561685_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563894_at2 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 234253 - -
          ABI Human Genome Survey Microarray Version 2.0 235129 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1696600 AL832737.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1696600 BC093745.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1696600 BC111990.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1696600 AL832737.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1696600 BC093745.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1696600 BC111990.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1696600 AL832737.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1696600 BC093745.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1696600 BC111990.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1696600 AL832737.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1696600 BC093745.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1696600 BC111990.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1696600 AL832737.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1696600 BC093745.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1696600 BC111990.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1696600 AL832737.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1696600 BC093745.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1696600 BC111990.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1696600 AL832737.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1696600 BC093745.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1696600 BC111990.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1696600 AL832737.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1696600 BC093745.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1696600 BC111990.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline