Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00330020_s1
          See other LOC157931 GE Assays ›
          Gene Symbol:
          LOC157931
          Gene Name:
          uncharacterized LOC157931
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00330020_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC157931
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK057274.1 - 1 252 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001984 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001943 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001975 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001925 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002020 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001968 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001970 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001972 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16274175 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001926 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001964 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001973 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001935 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001971 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001947 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001967 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16274173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07001993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07002010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27351598 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 92096 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 12132 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3823466 - -
          Agilent Human Genome CGH Microarray 44B A_14_P122597 - -
          Agilent Human Genome CGH Microarray 44A A_14_P122597 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 13482 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3823466 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P122597 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P122597 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P122597 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 147184 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 23325 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P122597 - -
          Agilent Human 1B Microarray G4111A A_32_P777067 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P122597 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561990_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0900009142.hg.1 ENST00000625008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009142.hg.1 ENST00000625008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009142.hg.1 ENST00000625008.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009142.hg.1 ENST00000625008.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009142.hg.1 ENST00000625008.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009142.hg.1 ENST00000625008.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009142.hg.1 ENST00000625008.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009142.hg.1 ENST00000625008.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1561990_at - -
          Human Genome U133 Plus 2.0 Array 1561990_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561990_at9 - -
          Human Genome U95 Set 42104_at - -
          Human Genome U95 Set 42104_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1841512 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 130341 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_74342 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1901656 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1901656 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1841512 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline