| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009072.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009072.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009072.hg.1 |
TC12000706.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009075.hg.1 |
ENST00000359455.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009075.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009075.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009075.hg.1 |
TC12000706.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009074.hg.1 |
ENST00000359455.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009074.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009074.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009074.hg.1 |
TC12000706.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008413.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200161742.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200161742.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200161742.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200161742.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200161742.hg.1 |
TC1200008413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200161740.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200161740.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200161740.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200161740.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200161740.hg.1 |
TC1200008413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200161743.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200161743.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200161743.hg.1 |
ENST00000546725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200161743.hg.1 |
NR_038868.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200161743.hg.1 |
TC1200008413.hg.1 |
Clariom D TC |