Search
Search
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319209.1 | NP_001306138.1 | 3 - 4 | 349 | 68 | |
| NM_002722.4 | NP_002713.1 | 3 - 4 | 308 | 68 | ||
| XM_011524978.2 | XP_011523280.1 | 3 - 4 | 462 | 68 | ||
| GenBank mRNA | BC032225.2 | - | 261 | 5229163 | 68 | |
| BC040033.2 | - | 2 - 3 | 261 | 5747221 | 68 | |
| BP388331.1 | - | 2 - 3 | 185 | 68 | ||
| BP394217.1 | - | 3 - 4 | 351 | 68 | ||
| X00491.1 | - | 2 - 3 | 275 | 68 | ||
Set Membership: |
|