| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019900.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019900.hg.1 |
ENST00000368407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019900.hg.1 |
ENST00000469878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019900.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019900.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019900.hg.1 |
TC01001288.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019903.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019903.hg.1 |
ENST00000368407.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019903.hg.1 |
ENST00000469878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019903.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019903.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019903.hg.1 |
TC01001288.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010111.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173656.hg.1 |
TC0100010111.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173655.hg.1 |
TC0100010111.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000368406.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000368407.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000469878.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000474413.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
ENST00000497282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
NM_004428.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
NM_182685.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173659.hg.1 |
TC0100010111.hg.1 |
Clariom D TC |