Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00358903_g1
          See other YBX1 GE Assays ›
          Gene Symbol:
          YBX1
          Gene Name:
          Y-box binding protein 1
          Gene Aliases:
          BP-8, CBF-A, CSDA2, CSDB, DBPB, EFI-A, MDR-NF1, NSEP-1, NSEP1, YB-1, YB1
          Chromosome Location:
          Chr.1: 42682235 - 42703803 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00358903_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: YBX1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004559.4 NP_004550.2 2 - 3 555 148
          GenBank mRNA AK309996.1 - 2 - 3 192 148
          BC000064.2 - 2 - 3 342 148
          BC002411.2 - 2 - 3 338 2961659 148
          BC010430.1 - 2 - 3 337 3506647 148
          BC015208.1 - 2 - 3 287 3914485 148
          BC018393.1 - 2 - 3 322 148
          BC038384.1 - 2 - 3 344 3449398 148
          BC065571.1 - 2 - 3 362 6171221 148
          BC070084.1 - 2 - 3 362 5272578 148
          BC071708.1 - 2 - 3 347 4361396 148
          BC072014.1 - 2 - 3 347 148
          BC090038.1 - 2 - 3 347 6526492 148
          BC098435.1 - 2 - 3 353 4859074 148
          BC106045.1 - 2 - 3 349 6062615 148
          CN353145.1 - 2 - 3 404 148
          J03827.1 - 2 - 3 350 148
          L28809.1 - 2 - 3 305 148
          M24070.1 - 2 - 3 344 148
          M83234.1 - 1 449 148
          M85234.1 - 1 455 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100008025.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene YBX1

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of cell division
          CRD-mediated mRNA stabilization
          protein localization to cytoplasmic stress granule
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          RNA binding
          protein binding
          poly(A) RNA binding
          GTPase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100156580.hg.1 TC0100008025.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 163592 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline