Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00360235_m1
          See other COA3 GE Assays ›
          Gene Symbol:
          COA3
          Gene Name:
          cytochrome c oxidase assembly factor 3
          Gene Aliases:
          CCDC56, HSPC009, MITRAC12
          Chromosome Location:
          Chr.17: 42797618 - 42798725 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00360235_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: COA3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001040431.2 NP_001035521.1 1 - 2 258 90
          GenBank mRNA AF070665.1 - 1 - 2 233 90
          AK225611.1 - 1 - 2 237 90
          AK290863.1 - 1 - 2 224 90
          AL524532.3 - 1 - 2 258 90
          AU122748.1 - 1 - 2 224 90
          BC002698.2 - 1 - 2 219 3609536 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010747.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001544.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial respiratory chain complex IV assembly
          positive regulation of mitochondrial translation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10328523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10328518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30901217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10328527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30901219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30901221 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10328521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10328531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10328529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30901214 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 748407 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P356453 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P100802 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P356453 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17598 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 36424 - -
          Agilent Human Genome CGH Microarray 44B A_14_P124991 - -
          Agilent Human Genome CGH Microarray 44A A_14_P124991 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 17048 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P20868113 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P356453 - -
          SurePrint G3 Human GE Microarray G4851A 3xSLv1 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P356453 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30901219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10328527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10328521 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30901221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30901214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10328531 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10328523 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10328523 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30901214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10328527 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30901219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30901221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10328531 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10328521 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P100802 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P356453 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P124991 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P124991 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P124991 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P356453 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P100802 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 89320 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2507206 - -
          Agilent Human 2 cDNA Microarray (G4101A) 2507206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328462 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328528 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328464 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328454 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16983886 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328521 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328524 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328530 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328519 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328451 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328456 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328458 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328523 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328460 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10328466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001040431_at - -
          Affymetrix Human Gene 1.0 ST Array 8015714 - -
          Affymetrix Human Gene 1.0 ST Array 8015713 - -
          Affymetrix Human Gene 1.1 ST Array 8015712 - -
          Affymetrix HT HG-U133+ PM Array Plate 236998_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 218026_PM_at - -
          Affymetrix Human Genome U219 Array 11761716_PM_at - -
          Affymetrix Human Genome U219 Array 11761716_at - -
          Affymetrix Human Genome U219 Array 11744040_a_at - -
          Affymetrix Human Genome U219 Array 11744040_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR17020633.hg.1 ENST00000328434.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17020633.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17020633.hg.1 TC17001544.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17020634.hg.1 ENST00000328434.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17020634.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17020634.hg.1 TC17001544.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 ENST00000586680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 ENST00000586680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 ENST00000586680.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010747.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700185507.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700185507.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700185507.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700185507.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700185507.hg.1 TC1700010747.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 ENST00000586680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 ENST00000586680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 ENST00000586680.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Clariom D Assay TC1700010747.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700185508.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700185508.hg.1 ENST00000586680.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700185508.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700185508.hg.1 ENST00000328434.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1700185508.hg.1 ENST00000586680.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700185508.hg.1 NM_001040431.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700185508.hg.1 TC1700010747.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 236998_PM_at - -
          Human Genome U133 Plus 2.0 Array 236998_at - -
          Human Genome U133 Plus 2.0 Array 218026_at - -
          Human Genome U133 Plus 2.0 Array 218026_PM_at - -
          Human Genome U133A 2.0 Array 218026_at - -
          Human Genome U133A 2.0 Array 218026_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw547718 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236998_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 218026_at4 - -
          Human Genome U95 Set 44713_at - -
          Human Genome U95 Set 44713_PM_at - -
          Human Genome U95 Set 78634_PM_at - -
          Human Genome U95 Set 78634_at - -
          Human Genome U133 Set 218026_at - -
          Human Genome U133 Set 218026_PM_at - -
          Human Genome U133 Set 236998_at - -
          Human Genome U133 Set 236998_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008377 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008377 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 175899 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1789786 - -
          HumanHT-12 v4 Expression BeadChip ILMN_2064898 BC002698.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2064898 CR593427.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2064898 CR594541.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2064898 NM_001040431.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2064898 AF070665.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1789786 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2064898 BC002698.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2064898 CR593427.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2064898 CR594541.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2064898 NM_001040431.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2064898 AF070665.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2064898 BC002698.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2064898 CR593427.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2064898 CR594541.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2064898 NM_001040431.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2064898 AF070665.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2064898 BC002698.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2064898 CR593427.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2064898 CR594541.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2064898 NM_001040431.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2064898 AF070665.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1789786 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2064898 BC002698.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2064898 CR593427.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2064898 CR594541.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2064898 NM_001040431.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2064898 AF070665.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2064898 BC002698.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2064898 CR593427.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2064898 CR594541.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2064898 NM_001040431.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2064898 AF070665.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1789786 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2064898 BC002698.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2064898 CR593427.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2064898 CR594541.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2064898 NM_001040431.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2064898 AF070665.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2064898 BC002698.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2064898 CR593427.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2064898 CR594541.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2064898 NM_001040431.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2064898 AF070665.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline