| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000504015.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000504015.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000504015.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000504015.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026741.hg.1 |
ENST00000331204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026741.hg.1 |
ENST00000505490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026741.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026741.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026741.hg.1 |
TC17001962.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008071.hg.1 |
ENST00000403118.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008071.hg.1 |
ENST00000403118.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026744.hg.1 |
ENST00000331204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026744.hg.1 |
ENST00000505490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026744.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026744.hg.1 |
TC17001962.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026743.hg.1 |
ENST00000331204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026743.hg.1 |
ENST00000505490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026743.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026743.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17026743.hg.1 |
TC17001962.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000504015.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000504015.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000504015.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000504015.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012078.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008071.hg.1 |
ENST00000403118.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008071.hg.1 |
ENST00000403118.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
ENST00000511412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198850.hg.1 |
TC1700012078.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700198853.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198853.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198853.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198853.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198853.hg.1 |
TC1700012078.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700198852.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198852.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198852.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198852.hg.1 |
ENST00000505490.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198852.hg.1 |
ENST00000512673.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700198852.hg.1 |
NM_005782.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700198852.hg.1 |
TC1700012078.hg.1 |
Clariom D TC |