Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00364284_g1
          See other KRAS GE Assays ›
          Gene Symbol:
          KRAS
          Gene Name:
          KRAS proto-oncogene, GTPase
          Gene Aliases:
          'C-K-RAS, C-K-RAS, CFC2, K-RAS2A, K-RAS2B, K-RAS4A, K-RAS4B, K-Ras, K-Ras 2, KI-RAS, KRAS1, KRAS2, NS, NS3, OES, RALD, RASK2, c-Ki-ras, c-Ki-ras2
          Chromosome Location:
          Chr.12: 25205246 - 25250929 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00364284_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRAS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001369786.1 NP_001356715.1 3 - 4 467 111
          NM_001369787.1 NP_001356716.1 3 - 4 467 111
          NM_004985.5 NP_004976.2 3 - 4 480 111
          NM_033360.4 NP_203524.1 3 - 4 480 111
          XM_047428826.1 XP_047284782.1 3 - 4 467 111
          GenBank mRNA AF493917.1 - 2 - 3 290 111
          AK292510.1 - 3 - 4 443 111
          BC013572.2 - 3 - 4 437 111
          BT007153.1 - 2 - 3 290 111
          M35504.1 - 2 - 3 290 111
          M35505.1 - 2 - 3 290 111
          M54968.1 - 3 - 4 482 111
          MK570065.1 - 2 - 3 360 111
          MZ068307.1 - 2 - 3 267 111
          MZ068308.1 - 2 - 3 254 111
          MZ068309.1 - 2 - 3 267 111
          MZ068310.1 - 2 - 3 254 111
          OR946359.1 - 78 111
          PV477844.1 - 2 - 3 290 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010158.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001314.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KRAS

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          liver development
          signal transduction
          Ras protein signal transduction
          female pregnancy
          cell proliferation
          visual learning
          response to gravity
          gene expression
          positive regulation of gene expression
          glial cell proliferation
          Rac protein signal transduction
          cytokine-mediated signaling pathway
          forebrain astrocyte development
          actin cytoskeleton organization
          negative regulation of epithelial cell differentiation
          regulation of synaptic transmission, GABAergic
          positive regulation of Rac protein signal transduction
          response to isolation stress
          skeletal muscle cell differentiation
          negative regulation of neuron apoptotic process
          regulation of long-term neuronal synaptic plasticity
          homeostasis of number of cells within a tissue
          striated muscle cell differentiation
          response to glucocorticoid
          response to mineralocorticoid
          neuron apoptotic process
          myoblast proliferation
          cardiac muscle cell proliferation
          positive regulation of glial cell proliferation
          epithelial tube branching involved in lung morphogenesis
          Type I pneumocyte differentiation
          positive regulation of cellular senescence
          nucleotide binding
          GTPase activity
          obsolete small monomeric GTPase activity
          protein binding
          GTP binding
          hydrolase activity
          GMP binding
          GDP binding
          LRR domain binding
          identical protein binding
          protein anchor
          macromolecular complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12017257.hg.1 ENST00000256078.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12017257.hg.1 ENST00000311936.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12017257.hg.1 NM_004985.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12017257.hg.1 NM_033360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12017257.hg.1 TC12001314.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200176874.hg.1 TC1200010158.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000256078.8 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000311936.7 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000556131.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 ENST00000557334.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_004985.4 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010158.hg.1 NM_033360.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline