Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00365015_m1
          See other MIA2 GE Assays ›
          Gene Symbol:
          MIA2
          Gene Name:
          MIA SH3 domain ER export factor 2
          Gene Aliases:
          CTAGE5, MEA6, MGEA, MGEA11, MGEA6, TALI
          Chromosome Location:
          Chr.14: 39233915 - 39388522 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00365015_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001329214.4 NP_001316143.1 1 - 2 315 99
          NM_054024.4 NP_473365.3 1 - 2 315 99
          XM_024449592.2 XP_024305360.1 1 - 2 315 99
          XM_024449593.2 XP_024305361.1 1 - 2 315 99
          XM_024449594.2 XP_024305362.1 1 - 2 315 99
          XM_024449595.2 XP_024305363.1 1 - 2 315 99
          XM_024449596.2 XP_024305364.1 1 - 2 315 99
          XM_024449597.2 XP_024305365.1 1 - 2 315 99
          XM_047431398.1 XP_047287354.1 1 - 2 315 99
          XM_047431401.1 XP_047287357.1 1 - 2 315 99
          XM_047431402.1 XP_047287358.1 1 - 2 315 99
          GenBank mRNA AF390175.2 - 1 - 2 115 99
          AK314069.1 - 1 - 2 315 99
          BC130537.1 - 1 - 2 147 99
          BC130563.1 - 1 - 2 146 99
          BX495701.1 - 1 - 2 320 99
          BX648228.1 - 1 - 2 314 99
          KX388743.1 - 1 - 2 115 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400010604.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14002295.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried MicroArray Quality Control (MAQC) Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MIA2

          Gene Ontology Categories:

          Function(s) Process(es)

          ER to Golgi vesicle-mediated transport
          protein secretion
          protein exit from endoplasmic reticulum
          cargo loading into vesicle
          lipoprotein transport
          protein localization to endoplasmic reticulum exit site
          protein binding
          enzyme activator activity
          cargo receptor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14002655.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14002655.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14002655.hg.1 TC14002295.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR14002653.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14002653.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14002653.hg.1 TC14002295.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14002657.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14002657.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14002657.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14002657.hg.1 TC14002295.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR14002656.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14002656.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14002656.hg.1 NM_054024.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14002656.hg.1 TC14002295.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179288.hg.1 TC1400010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400179285.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179285.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179285.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179285.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179285.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179285.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179285.hg.1 TC1400010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179289.hg.1 TC1400010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010604.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179286.hg.1 TC1400010604.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 ENST00000280082.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 ENST00000555143.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 ENST00000556784.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 ENST00000557148.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 NM_054024.3 Refseq
          Affymetrix Human Clariom D Assay PSR1400179290.hg.1 TC1400010604.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 102347 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0